Product BDPH9C021C8C is not currently sold in your country.
Inchi Code
1S/C3H5N3O/c4-2-1-5-6-3(2)7/h1H,4H2,(H2,5,6,7)
Inchi Key
NATMWGYJBMRROQ-UHFFFAOYSA-N
Shipping Temperature
Normal
Purity
95%
Storage temp.
Keep in dark place, inert atmosphere, room temperature
Country of Origin
CN
Physical Form
Powder or crystals
Still not finding the right product?
Explore all of our products under 4-Amino-1H-pyrazol-3(2H)-one
hstatements
signalword
Warning
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
