Skip to Content
Merck

BDPH9C021CD0

Bide Pharmatech

H-Ser(tBu)-OH

Synonym(s):

(2S)-2-amino-3-tert-butoxypropanoic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Product BDPH9C021CD0 is not currently sold in your country.

About This Item

Linear Formula:
C7H15O3N1
CAS Number:
Molecular Weight:
161.2
MDL number:
Product BDPH9C021CD0 is not currently sold in your country.

Inchi Code

1S/C7H15NO3/c1-7(2,3)11-4-5(8)6(9)10/h5H,4,8H2,1-3H3,(H,9,10)/t5-/m0/s1

Inchi Key

DDCPKNYKNWXULB-YFKPBYRVSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Keep in dark place, inert atmosphere, room temperature

Country of Origin

CN

Physical Form

Form: Crystal - Powder / Colour: White - Almost white

Looking for similar products? Visit Product Comparison Guide

signalword

Warning

pictograms

Exclamation mark

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library