Skip to Content
Merck

BDPH9C021E01

Bide Pharmatech

bicyclo[3.1.0]hexan-2-one

Synonym(s):

bicyclo[3.1.0]hexan-2-one

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C6H8O1
CAS Number:
Molecular Weight:
96.13
MDL number:
Product BDPH9C021E01 is not currently sold in your country.


Inchi Code

1S/C6H8O/c7-6-2-1-4-3-5(4)6/h4-5H,1-3H2

Inchi Key

HFCMQKFXCMZZMZ-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

97%

Storage temp.

Inert atmosphere, 2-8C

Country of Origin

CN

Physical Form

Liquid



Still not finding the right product?

Explore all of our products under bicyclo[3.1.0]hexan-2-one


hstatements

signalword

Danger

pictograms

Flame


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library