Skip to Content
Merck

BDPH9C021E4B

Bide Pharmatech

4-Methoxy-1H-pyrrolo[2,3-c]pyridine

Synonym(s):

4-methoxy-1H-pyrrolo[2,3-c]pyridine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C8H8O1N2
CAS Number:
Molecular Weight:
148.16
MDL number:
Product BDPH9C021E4B is not currently sold in your country.


Inchi Code

1S/C8H8N2O/c1-11-8-5-9-4-7-6(8)2-3-10-7/h2-5,10H,1H3

Inchi Key

UPDSOLUKSUHRDC-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Sealed in dry, room temperature

Country of Origin

CN

Physical Form

Form: solid



Still not finding the right product?


hstatements

signalword

Warning

pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library