Skip to Content
Merck

BDPH9C021F2B

Bide Pharmatech

Methyl 4-(1H-indol-2-yl)benzoate

Synonym(s):

methyl 4-(1H-indol-2-yl)benzoate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
C16H13O2N1
CAS Number:
Molecular Weight:
251.28
MDL number:
Product BDPH9C021F2B is not currently sold in your country.


Inchi Code

1S/C16H13NO2/c1-19-16(18)12-8-6-11(7-9-12)15-10-13-4-2-3-5-14(13)17-15/h2-10,17H,1H3

Inchi Key

LXPCCNBTEKIKLX-UHFFFAOYSA-N

Shipping Temperature

Normal

Purity

95%

Storage temp.

Keep in dark place, sealed in dry, room temperature

Country of Origin

CN

Physical Form

Powder or crystals



Still not finding the right product?

Explore all of our products under Methyl 4-(1H-indol-2-yl)benzoate


hstatements

pcodes

signalword

Warning

pictograms

Exclamation mark


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library