Product BDPH9C021F48 is not currently sold in your country.
Inchi Code
1S/C44H54N7O7P/c1-30(2)51(31(3)4)59(56-26-12-24-45)58-38-27-40(50-25-23-37-41(50)47-43(48-42(37)52)46-29-49(5)6)57-39(38)28-55-44(32-13-10-9-11-14-32,33-15-19-35(53-7)20-16-33)34-17-21-36(54-8)22-18-34/h9-11,13-23,25,29-31,38-40H,12,26-28H2,1-8H3,(H,47,48,52)/b46-29+/t38-,39+,40+,59?/m0/s1
Inchi Key
HBENOWJVIKVUGF-OLQNVDSXSA-N
Shipping Temperature
Cold-chain transportation
Purity
95%
Storage temp.
Keep in dark place, inert atmosphere, store in freezer, under -20C
Country of Origin
CN
Physical Form
Powder or crystals
Still not finding the right product?
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
