Skip to Content
Merck

KEY128004154

Key Organics / BIONET

2-fluoro-6-(2,2,2-trifluoroethoxy)benzonitrile

Sign In to View Organizational & Contract Pricing

Select a Size

Product KEY128004154 is not currently sold in your country.

About This Item

CAS Number:
Molecular Weight:
219.14
MDL number:
Product KEY128004154 is not currently sold in your country.

mp

88 - 90

Iupac Name

2-fluoro-6-(2,2,2-trifluoroethoxy)benzonitrile

Inchi Code

1S/C9H5F4NO/c10-7-2-1-3-8(6(7)4-14)15-5-9(11,12)13/h1-3H,5H2

InChI key

KPUXDDKZMLVAEI-UHFFFAOYSA-N

Purity

95%

Country of Origin

GB

Physical Form

Solid

Product Link

https://www.keyorganics.net/sku/FA-0838

Melting Point

88 - 90

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Codes

Precautionary statements

MSDS

https://www.keyorganics.net/sds/FA-0838


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library