Skip to Content
Merck

219503

Cdk4 Inhibitor V

The Cdk4 Inhibitor V, also referenced under CAS 943746-57-4, controls the biological activity of Cdk4. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

Synonym(s):

Cdk4 Inhibitor V, 4-(((4-Hydroxy-5-propoxy-pyridin-2-ylmethyl)-amino)-methylene)-6-iodo-4H-isoquinoline-1,3-dione

Sign In to View Organizational & Contract Pricing.

Select a Size


About This Item

Empirical Formula (Hill Notation):
C19H18IN3O4
CAS Number:
Molecular Weight:
479.27
UNSPSC Code:
12352200
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Product Name

Cdk4 Inhibitor V, The Cdk4 Inhibitor V, also referenced under CAS 943746-57-4, controls the biological activity of Cdk4. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.

SMILES string

O=C(/C1=C\NCC2=CC(O)=C(OCCC)C=N2)NC(C3=C1C=C(I)C=C3)=O

assay

≥98% (HPLC)

form

powder

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

color

caramel

solubility

DMSO: 50 mg/mL

shipped in

ambient

storage temp.

2-8°C

Quality Level

Disclaimer

Toxicity: Regulatory Review (Z)

General description

A cell-permeable and metabolically stable (t1/2 >30 min at 37 °C in the presence of rat liver microsomes, NADPH, and UDPGA) pyridinylmethylamino-isoquinoline dione that acts as a potent, ATP-binding site-targeting inhibitor against Cdk4/D1 kinase activity (IC50 = 30 nM in cell-free Rb c-terminal fragment phosphorylation assays) and an effective antiproliferative agent against HCT116 colorectal and MCF-7 breast cancer cultures (IC50 = 0.73 and 0.88 µM, respectively), while affecting Cdk2/E and Cdk1/B kinase activities only at much higher concentrations (IC50 = 1.6 and 25.3 µM, respectively).

Other Notes

Tsou, H.R., et al. 2009. J. Med. Chem.52, 2289.

Packaging

Packaged under inert gas

Preparation Note

Following reconstitution, aliquot and freeze (-20°C). Stock solutions are stable for up to 6 months at -20°C.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class

11 - Combustible Solids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

New Products: Antibodies, Assays, Small Molecules, Inhibitors, and Proteins

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service