Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|---|---|
Estimated to ship on26 October 2026fromAreál Kühne+Nagel spol. s r.o. | 4 170,00 CZK |
About This Item
NACRES:
NA.31
UNSPSC Code:
12352200
Quality Segment
1 of 1
This Item | |||
|---|---|---|---|
| description - | description - | description - | description - |
| Quality Level 100 | Quality Level 100 | Quality Level 100 | Quality Level 200 |
Storage Class
12 - Non Combustible Liquids
WGK
WGK 2
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



