Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
biological source
rabbit
conjugate
unconjugated
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
polyclonal
purified by
affinity chromatography
species reactivity
human, rat
species reactivity (predicted by homology)
mouse (based on 70% sequence homology)
technique(s)
immunohistochemistry: suitable (paraffin), western blot: suitable
NCBI accession no.
UniProt accession no.
shipped in
wet ice
target post-translational modification
unmodified
Gene Information
human ... PATE1(160065)
General description
Immunogen
Application
Apoptosis & Cancer
Apoptosis - Additional
Physical form
Preparation Note
Analysis Note
Western Blotting Analysis: A 1:500 dilution of this antibody detected PATE1 in 10 µg of rat testis tissue lysate.
Other Notes
Disclaimer
1 of 1
This Item | |||
|---|---|---|---|
| description from rabbit, purified by affinity chromatography | description from rabbit, purified by affinity chromatography | description Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody | description - |
| antibody form affinity isolated antibody | antibody form affinity isolated antibody | antibody form affinity isolated antibody | antibody form affinity purified immunoglobulin |
| Quality Level 100 | Quality Level 100 | Quality Level - | Quality Level 300 |
| biological source rabbit | biological source rabbit | biological source rabbit | biological source - |
| conjugate unconjugated | conjugate unconjugated | conjugate unconjugated | conjugate unconjugated |
| UniProt accession no. | UniProt accession no. | UniProt accession no. - | UniProt accession no. - |
| shipped in wet ice | shipped in wet ice | shipped in wet ice | shipped in - |
Still not finding the right product?
Explore all of our products under Anti-PATE1 Antibody
— or —
Try our Product Selector Tool to narrow your options
Storage Class
12 - Non Combustible Liquids
WGK
WGK 1
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



