Select a Size
CZK 4,040.00
CZK 8,470.00
About This Item
Skip To
biological source
rabbit
Quality Level
conjugate
unconjugated
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
polyclonal
species reactivity
human
packaging
antibody small pack of 25 μL
technique(s)
immunohistochemistry: suitable (paraffin)
western blot: suitable
NCBI accession no.
UniProt accession no.
target post-translational modification
unmodified
Gene Information
human ... ZBTB38(253461)
1 of 4
This Item | SAB4503733 | AV31874 | HPA020306 |
|---|---|---|---|
| biological source rabbit | biological source rabbit | biological source rabbit | biological source rabbit |
| Quality Level 100 | Quality Level 100 | Quality Level 100 | Quality Level 100 |
| antibody form affinity isolated antibody | antibody form affinity isolated antibody | antibody form affinity isolated antibody | antibody form affinity isolated antibody |
| species reactivity human | species reactivity mouse, human | species reactivity rat, human, bovine, guinea pig, rabbit | species reactivity human |
| technique(s) immunohistochemistry: suitable (paraffin), western blot: suitable | technique(s) ELISA: 1:10000, immunohistochemistry: 1:50-1:100 | technique(s) western blot: suitable | technique(s) immunofluorescence: 0.25-2 μg/mL, immunohistochemistry: 1:200-1:500 |
| Gene Information human ... ZBTB38(253461) | Gene Information human ... ZBTB40(9923) | Gene Information human ... ZBTB32(27033) | Gene Information human ... ZBTB39(9880) |
General description
Immunogen
Application
Epigenetics & Nuclear Function
Biochem/physiol Actions
Physical form
Preparation Note
Analysis Note
Western Blotting Analysis: A 1:500 dilution of this antibody detected ZBTB38 in HeLa cell nuclear extract.
Other Notes
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
12 - Non Combustible Liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Active Filters
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service


