Skip to Content
Merck

MX0SP10FS36AS

Millistak+® HC Pro Pod Depth Filter

new

X0SP, filtration area 1.1 m2, Process Scale in APAC format Optimized Packaging 36/PK

Synonym(s):

1.1m2 surface aera, Millistak+® HC Pro Pod Depth Filter, Optimized Packaging, Process Scale Pod

Sign In to View Organizational & Contract Pricing

Select a Size


Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

material

acetal (polypropylene) housing
filter aid (Synthetic filter media)

Quality Level

product line

Millistak+®

feature

hydrophilic

filtration area

1.1 m2

impurities

(LAL test)

input

liquid

fitting

fitting Flat Seal

application(s)

pharma/biopharma processes

code

X0SP

shipped in

ambient

General description

To improve sustainability of the packaging and shipping of filter products, Optimized Pack solutions for Millistak+® HC Pro Process Scale Pod filters have been developed to optimize transport and receiving processes as well as to reduce waste.

Application

Process Scale manufacturing Secondary clarification post depth filtration or centrifugation. Effective removal of aggregates post viral inactivation or before virus filtration. Can also be utilized in downstream processing steps to protect chromatography columns.

Features and Benefits

The key benefits are:
  • 42% average reduction in corrugated packaging material per product.
  • 29% decrease in the number of pallets further reducing energy use and emissions.
  • 75% reduction in operator time to open and manage the product and packaging.

Packaging

36/pack Optimized packaging configuration includes 36 pods distributed in 3 boxes on one APAC standardized pallet (pallet size: 1100*1100mm).

Other Notes

Integrity is maintained after 1 autoclave cycle of 60 minutes at 123 °C. Filtration performance may be impacted post autoclave. Recommended for post-use decontamination only

Legal Information

MILLISTAK+ is a registered trademark of Merck KGaA, Darmstadt, Germany

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service