Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
Quality Segment
grade
Molecular Biology
form
liquid
mol wt
547.55 g/mol
storage condition
(Cold storage (2-8°C))
technique(s)
PCR: suitable, qPCR: suitable
impurities
DNA, RNA, Dnase, Rnase, Nickase free
General description
Designed for the most demanding molecular applications, our ROX Reference Dye is prepared to the highest standards of purity, ensuring a DNA-free formulation that eliminates the risk of background contamination in sensitive qPCR assays.
Application
Features and Benefits
- DNA-free
- DNase-free, RNase-free, and nickase-free
- Enhanced change control and quality agreement
- Ready-to-use 100× solution – convenient and consistent, with no preparation required
- Excitation/Emission: 586 nm / 605 nm – compatible with standard ROX channel instrument settings
- Broad chemistry compatibility – works with SYBR Green, molecular probes, and dual-labeled probe chemistries
- Lot-to-lot consistency – rigorous quality control ensures reproducibility across experiments
- Suitable for further manufacture – meets the quality and regulatory requirements of highly regulated industries
Other Notes
- Uncompromising Purity-
- Our Ultra pure & DNA-free ROX Reference Dye is manufactured to the highest standards, ensuring an ultra-pure formulation free from contaminants that could interfere with sensitive qPCR reactions. Every batch undergoes stringent quality control testing to guarantee consistent dye concentration and fluorescence output.
- DNA-Free- Contaminating DNA in a passive reference dye can introduce false signals and compromise the integrity of your qPCR data. Our DNA-free certification gives you full confidence that your normalization standard will never be a source of spurious amplification or background noise.
- Ready-to-Use Convenience- Supplied as a pre-formulated 100× solution, our ROX Reference Dye eliminates the need for time-consuming preparation or dilution optimization. Simply add the appropriate volume to your mastermix and proceed directly to your experiment, reducing hands-on time and the risk of preparation errors.
- Broad Platform and Chemistry Compatibility-Our Ultra pure & DNA-free ROX Reference Dye is compatible with a wide range of real-time PCR instruments and detection chemistries, including:
- SYBR Green intercalating dye assays
- Molecular probe chemistries
- Dual-labeled probe (e.g., TaqMan) chemistries
- Consistent, Reproducible Normalization- Lot-to-lot variability in a passive reference dye can introduce systematic errors across experiments. Our rigorous manufacturing and quality control processes ensure that every lot delivers the same fluorescence intensity and spectral profile, giving you a reliable normalization baseline experiment after experiment.
- Trusted Across Diverse Applications- Our Ultra pure & DNA-free ROX Reference Dye has been validated for use in a broad range of real-world qPCR applications, giving you confidence that it will perform reliably in your specific workflow.
1 of 1
This Item | |||
|---|---|---|---|
| description Ultra pure, Molecular Biology grade reference dye for Quantitative PCR | description Ultra pure, Molecular Biology grade, 1M | description 100 ×, solution | description Combined Ultra pure BST-RT polymerase, Ultra pure BST-RT Reaction Buffer and Magnesium sulfate |
| grade Molecular Biology | grade Molecular Biology | grade - | grade Molecular Biology |
| technique(s) qPCR: suitable | technique(s) qPCR: suitable | technique(s) PCR: suitable, whole genome amplification: suitable | technique(s) cDNA synthesis: suitable, RT-qPCR: suitable (Isothermal amplification) |
| Quality Level 400 | Quality Level 400 | Quality Level 300 | Quality Level 400 |
| form liquid | form liquid | form solution | form liquid |
| storage condition (Cold storage (2-8°C)) | storage condition room temperature (2-30 °C) | storage condition - | storage condition avoid repeated freeze/thaw cycles |
| impurities DNA, RNA, Dnase, Rnase, Nickase free | impurities DNA, RNA, Dnase, Rnase, Nickase free | impurities - | impurities (DNase free, RNasefree, DNA free, NICKase free) |
Storage Class
10 - Combustible liquids
WGK
WGK 2
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.


