Skip to Content
Merck

Skip To

A9571

MilliShot® Antibiotic Antimycotic Solution

0.2 μm filtered, BioReagent, suitable for cell culture

Synonym(s):

Penicillin-Streptomycin-Amphotericin B Suspension

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
20 x 5 mL
Please contact Customer Service for Availability
1 340,00 CZK

About This Item

NACRES:
NA.46
eCl@ss:
32035490

1 340,00 CZK


Please contact Customer Service for Availability

​
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

yeast

Quality Segment

sterility

0.2 μm filtered

product line

BioReagent

form

liquid

shelf life

2 yr

shipped in

dry ice

storage temp.

-10 to -25°C

General description

Antibiotic Antimycotic Solution is formulated to contain 10,000 units penicillin, 10 mg streptomycin and 25μg Amphotericin B per mL, and is solubilized in a proprietary citrate buffer. It is recommended for use in cell culture applications at 10mL/L. Antibioticantimycotic solution is generally used to prevent the growth of harmful microorganisms including bacteria and fungi in cell culture media.

Application

Antibiotic antimycotic solution has been used:
  • As a supplement in different types of cell culture media.
  • As a component of phosphate buffer saline (PBS) sterile solution to prevent contamination of periodontal ligament fibroblasts (PDLF) tissues that are isolated and cultured in the experiments.
  • As a component of storage solution for preserving tracheal segments harvested for subsequent experimental investigation.

Biochem/physiol Actions

Mode of Action: Penicillin acts by inhibiting bacterial cell-wall synthesis. Streptomycin inhibits prokaryote protein synthesis by preventing the transition from initiation complex to chain-elongating ribosome and causes miscoding. Amphotericin B interferes with fungal membrane permeability by forming channels in the membranes and causing small molecules to leak out.
Antimicrobial spectrum: Includes Gram-negative bacteria, Gram-positive bacteria, fungi and yeasts.

Legal Information

MILLISHOT is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
A5955P9080A9570
description

0.2 μm filtered, BioReagent, suitable for cell culture

description

with 10,000 units penicillin, 10 mg streptomycin and 25 μg amphotericin B per mL, 0.1 μm filtered, BioReagent, suitable for cell culture

description

0.2 μm filtered, BioReagent, suitable for cell culture

description

250 μg/mL in deionized water, 0.2 μm filtered, BioReagent, suitable for cell culture

product line

BioReagent

product line

BioReagent

product line

BioReagent

product line

BioReagent

Quality Level

300

Quality Level

200

Quality Level

300

Quality Level

300

form

liquid

form

suspension

form

liquid

form

liquid

storage temp.

-10 to -25°C

storage temp.

−20°C

storage temp.

-10 to -25°C

storage temp.

-10 to -25°C

biological source

yeast

biological source

-

biological source

yeast

biological source

yeast

sterility

0.2 μm filtered

sterility

0.1 μm filtered

sterility

0.2 μm filtered

sterility

0.2 μm filtered


Storage Class

12 - Non Combustible Liquids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library