Skip to Content
Merck

ES-R1-EYFP 7AC5/EYFP Mouse Embryo, ECACC 07072003

NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.

Synonym(s):

B5/EGFP, ES-R 1, ES-R-1, ESR-1, ESR1

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41106514
eCl@ss:
32160903
Biological source:
mouse embryo
Growth mode:
Adherent
Karyotype:
Not specified
Morphology:
Adherent monolayer of spheroidal cells on feeder layer of mouse primary embryonic fibroblasts
Manufacturer/tradename:
ECACC
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

ES-R1-EYFP 7AC5/EYFP, NOTE: Both the cell line and DNA from the cell line may be available for this product. Please choose -1VL or VIAL for cells, or -DNA-5UG for DNA.

biological source

mouse embryo

manufacturer/tradename

ECACC

packaging

tube of 5 μg 07072003-DNA-5UG, pkg of vial of cells 07072003-1VL

growth mode

Adherent

karyotype

Not specified

morphology

Adherent monolayer of spheroidal cells on feeder layer of mouse primary embryonic fibroblasts

products

Not specified

receptors

Not specified

technique(s)

cell culture | mammalian: suitable

shipped in

dry ice

storage temp.

-140 to -196°C

Biochem/physiol Actions

Mouse embryonic stem cell, YFP expression
Pluripotent mouse embryonic stem cell line expressing EYFP. This yellow fluorescent variant was generated by the random integration of EYFP transgenes into ES-R1 (ECACC 07072001) using co-electroporation with a circluar selectable marker containing vector pPGK Puro. The vector is driven by a CMV immediate early enhancer coupled to the chicken beta-actin promoter and first intron.

Preparation Note

Embryonic stem (ES) cells require the use of mitotically inactivated feeder cells to support the growth of stem cells in the undifferentiated state. Mouse embryonic fibroblasts, STO (Sigma product number 86032003) or SNL 76/7 (Sigma product number 07032801) can be used. At ECACC plastic ware is pre-coated with gelatine prior to plating feeder cells. Porcine gelatine (Sigma product numbe G1890) is dissolved in sterile water (0.5 g/500ml) at 56 °C. The 0.1% solution is sterilized by filtration (0.22 μm). Add 0.1% gelatine to plastic ware to cover bottom, and incubate for 20 minutes at room temperature (RT). Remove gelatine, wash with PBS once and replace with appropriate culture medium. The flask/dish must not be allowed to dry out. Feeder layers are prepared on the gelatinized flasks at least 24 hours in advance of being required. An ampoule is thawed in 37 °C water bath and the contents quickly transferred to a 15 ml centrifuge tube. MEF medium is added drop wise to 5 ml. Cells are centrifuged at 150 x g for 5 minutes at RT. Cells are resuspended in 5 ml of MEF medium. Cells are counted and added to flasks containing the correct medium at 1-3 x 104 cells/cm2. An ampoule of ES cells is thawed in 37 °C water bath and the contents quickly transferred to a 15 ml centrifuge tube. KSR medium is added drop wise to 5 ml. Cells are centrifuged at 150 x g for 5 minutes. Cells are resuspended in 5 ml of KSR medium. The prepared feeder flask is washed once with PBS and KSR medium added. ES cells should be plated at 4-5 x 104 cells/cm2.  Cultures must be incubated in a humidified 5% CO2/95% air incubator at 37 °C. A 100% media change must be performed every day and cells passaged every 2-3 days. Colonies must not be allowed to touch each other as overgrowth will result in differentiation.
MEF medium consists of Advanced DMEM/F12, 10% FBS / FCS (F2442), 2 mM L-Glutamine (G7513) and 0.1 mM 2-Mercaptoethanol (M6250). KSR medium consists of DMEM, 20% Serum Replacer, 2 mM L-Glutamine (G7513), NEAA (M7145), 0.1 mM 2-Mercaptoethanol (M6250) and 1000 Units/ml LIF.

Other Notes

Additional freight & handling charges may be applicable for Asia-Pacific shipments. Please check with your local Customer Service representative for more information.
This cell line is part of the European Collection of Authenticated Cell Cultures (ECACC), an international repository managed by the United Kingdom Health Security Agency (UKHSA). Use of this product by commercial organizations may require a license agreement. Commercial customers will be contacted by cellscreening@merckgroup.com to provide further information after the order request is submitted. Any other use of these products is prohibited without the express written permission of UKHSA. Inquiries regarding authorized use of this cell line or its genomic DNA may be directed to culturecollections@ukhsa.gov.uk.

Disclaimer

This cell line has special release conditions: Commercial organisations are required to complete the ′Cell Line Release Authorisation for Research Use in Commercial Organisations′ release conditions form.




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library