Skip to Content
Merck

EHNC005481

MISSION® esiRNA

targeting human AC079766

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.51
UNSPSC Code:
41105324
eCl@ss:
32119207
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GTTTACGGCGGCAAGAGAGCTCAATTAATTTCTGAGGGAGGACGGCAGGATGCGACCCCTTGTGCGTGGCCTAGGAGGAACTGTCCCTGTGTGAATTTATGTTCTGGTAATTTAAAAAGATTTATATTAAGCTTTTCTTTCATTTTGAGAGTAGATTAAAATTCTAATGGAAACCATTTGTATGCATTTGTTGAATTGAAGAGTATTTATGGCTGTGTAAAAAAAATGCTGCAAAATTCTGCCGATAATAAAGTCAGTGAAGCAAGAATAAGTGTAAAAACAGGAAGGAAATACGCGA

GenBank accession no.

shipped in

ambient

storage temp.

−20°C

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services
MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany


Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable

WGK

WGK 1

Storage Class

12 - Non Combustible Liquids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library