Skip to Content
Merck

Skip To

EHNC011171

MISSION® esiRNA

targeting human AC010183

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
20 μg
Please contact Customer Service for Availability
5 690,00 CZK
50 μg
Please contact Customer Service for Availability
10 300,00 CZK

About This Item

NACRES:
NA.51
UNSPSC Code:
41105324
eCl@ss:
32119207

5 690,00 CZK


Please contact Customer Service for Availability

​
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

GCTGACAGCCCTCTCAATTCTGAGATTTGTTATTCAGTCTTTTCCCAATAGGATCTACAAAGCACCTCTTGACCATTTATCATGCATGGAACCAACATGTCAAATTCCTCAATAACTGATTCCCGCCTAGGATCACACTCATTGACTAGCTCCGCCAATTCTTACAATTCATTCAGGCTCTGCTACAAGCATACAGCTTTGCTAGAAGTAGATTTCTTCTAGGAAATTTCATGCTACTGAGAGGTTGTGTGGCAGAGCCATGTTGCCATTCTTCCAGCATGACTTGATAGCTAAGTTGAACACTATCCCAAGGAGATGCCTCCTTGACAACTTACACTGCAAAGTGAATCATAACCCTGACCCTGGAGACTGCTATAACGGGCTCCTGTGAGAGTTCTATTTGCAGAACCCAGAATTCACAACGG

GenBank accession no.

shipped in

ambient

storage temp.

−20°C

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services
MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
EHNC016061EHNC001151EHNC016621
description

targeting human AC010183

description

targeting human AC098932

description

targeting human LINC01320

description

targeting human AL138963

product line

MISSION®

product line

MISSION®

product line

MISSION®

product line

MISSION®

esiRNA cDNA target sequence

GCTGACAGCCCTCTCAATTCTGAGATTTGTTATTCAGTCTTTTCCCAATAGGATCTACAAAGCACCTCTTGACCATTTATCATGCATGGAACCAACATGTCAAATTCCTCAATAACTGATTCCCGCCTAGGATCACACTCATTGACTAGCTCCGCCAATTCTTACAATTCATTCAGGCTCTGCTACAAGCATACAGCTTTGCTAGAAGTAGATTTCTTCTAGGAAATTTCATGCTACTGAGAGGTTGTGTGGCAGAGCCATGTTGCCATTCTTCCAGCATGACTTGATAGCTAAGTTGAACACTATCCCAAGGAGATGCCTCCTTGACAACTTACACTGCAAAGTGAATCATAACCCTGACCCTGGAGACTGCTATAACGGGCTCCTGTGAGAGTTCTATTTGCAGAACCCAGAATTCACAACGG

esiRNA cDNA target sequence

TGAAGAGGGCTGTGGAAACTTAAGTCATAGACATGCTTCAGTGAGAGGATGAGGATTTGCCCAGATGGGAAGCTTCAAGTGCAGAGGCACTTGTACATCTACTTATCTTGAGTCTCAATGATGCTGTTTATGAGTGAGCTATTCAAATGGAGGAAGAAACCCACGGAGAAGGAGAAATAACAGGTCGGAGTGATTATGTACAGAAGGATAGGCTCTAGCAGGAGTCCAGTGGGAAAGAAGGAAGGGAATACAATTAAGAATTATTAATGGCAGATCTCTAATGATTTCACTGGATCCAAAGGAACAGGGCAATACTGAGTAAGAAACACTTGATATACGAAAAAAGCGTGAGTGCGAAGGAAAATACATCATTCTACAGATGTTGACGGACTCTGACGGTGGGAAAGCGTCCCTCAAGATTG

esiRNA cDNA target sequence

GGCCATGGCTTCAACTAGACTCCACATCTTGAATCACAGAACTCAACGAATGGCATCAACTACAAGCTGGTATTCTGCTATACATTTAAGAGGCCATTTTAATATTCTATTAGTCTAGCAGCTTCTAAGTGATGAGGGATATGGGGGGACTGGAGGATCCCGTGCTCACTTCCTTTTCTGTAAAAGGAGCTCTTTGATCCAAGGTGACATGGTATGGGATCCAATGACAATAAATCAGTTATTCTGTGAACCCTGATCAAAACTTGACTAAACTTTTGACAGTTAGGTTTTTTCCTGACCACAGACTCCTCACCTTTCTTTTCTTAGAAAATTTACATTGGAAAACAGATGTAAATTTTCTCCAGCCTCTTGCAAGGTTTACAGTCCAGGACTTTGAGGTCCTGGGAGTCATCCTTTTGAAATACAATCATCAAAGAAGATAGTGCTCCAATCTCCTAGTCTCTGTGGGAGGGTTGGAACCTAACTTCAGTAAACTAATCACCAATTAGCAAACACAGAGGGGCTGATCAT

esiRNA cDNA target sequence

TCTGCATCAATTTGGCAGACTTACAAGAAAAAAAGGCATTCTTTCCATTTTGGAAGAAGGCATGTGGAATTAGTACTATGCAGTGGTGGTATTTTGAAATCTGGGTTCTAATAAAAGCCCAACCACTTGCTGTGAGAACTCCAGCATCTTAACCTCTCTAAGCCTCAGTATCTCCATCTAGAAAACAGAAACTCATACTTACCTCATCGGCTGATTTGGTAAGTAGAGGCTATCATAGGAGTGCCTTTACCATGAGAAATTCTTACAATACTGGTCTCTACTCTGAATCAGTGCAAAATACAGGATGCCTGTTCACATTCTCTGACTTGCAAAATACAGGATGCCTGTTCACATTCTCTGACTTTTGTGTAGCCTTGGGAAACTGCATCATGAATATGTTTTAGGGGTCAGGCG

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

form

lyophilized powder

form

lyophilized powder

form

lyophilized powder

form

lyophilized powder

GenBank accession no.

AC010183

GenBank accession no.

AC098932

GenBank accession no.

AC073218

GenBank accession no.

AL138963

shipped in

ambient

shipped in

ambient

shipped in

ambient

shipped in

ambient


Storage Class

10 - Combustible liquids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters