Skip to Content
Merck

Skip To

EHNC012631

MISSION® esiRNA

targeting human AC007590

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
20 μg
Please contact Customer Service for Availability
5 690,00 CZK
50 μg
Please contact Customer Service for Availability
10 100,00 CZK

About This Item

NACRES:
NA.51
UNSPSC Code:
41105324
eCl@ss:
32119207

5 690,00 CZK


Please contact Customer Service for Availability

​
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

TGGAGAAGACCAGGAGAGGAAGCAGAAGACCCTCCTCAAAGAAGCAGCCTTCACCTCCAGACCTCCCAGCGGGACTTAGACAATGGGGTTAGGCAGTCACATGTCTGCACGTGGAAACTTTCTGGGACTGTGCTTGGATCCTGGCTCTGCCTTTTACTAGCCATGTGACCTTGGGAAAATCACTTGACTTCTCCGTGCAGGATTCCTCATCTGTAAAAATGGTCTAAGATTAGGAGCCACCTCAAGGTTG

GenBank accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services
MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
EMNC010311EHNC002781EHNC018891
description

targeting human AC007590

description

targeting mouse Scn2a

description

targeting human AC009330

description

targeting human AC107223

Gene Information

human ... LOC100507420(100507420)

Gene Information

mouse ... SCN2A(110876)

Gene Information

-

Gene Information

-

product line

MISSION®

product line

MISSION®

product line

MISSION®

product line

MISSION®

esiRNA cDNA target sequence

TGGAGAAGACCAGGAGAGGAAGCAGAAGACCCTCCTCAAAGAAGCAGCCTTCACCTCCAGACCTCCCAGCGGGACTTAGACAATGGGGTTAGGCAGTCACATGTCTGCACGTGGAAACTTTCTGGGACTGTGCTTGGATCCTGGCTCTGCCTTTTACTAGCCATGTGACCTTGGGAAAATCACTTGACTTCTCCGTGCAGGATTCCTCATCTGTAAAAATGGTCTAAGATTAGGAGCCACCTCAAGGTTG

esiRNA cDNA target sequence

CCTTGTGCTCCTGAATGACAAACAGCCTTATAGAAGCTCATTGTTCTACATATTCTATTCTATAGAATGGTCTGTGTGCATATAGAGAGAGTAATTTGTGAATACTTAGATGGTTATACAGATTATATAATACCTCTTCAATATCTGGAGAAAAGAAGGGCTTTAACATGTGTCATTTCTACATAAATACTTGTGGAATATCATATCTTTACTGAAGAGTGGTTCATGGTAGGAATTGAAAAGTAATTACAGGGACTAAGAGATGGCTCTGTGGCTAACAGTGAGCACTGCTCTTGCAGAGGGACCTGCATTCAGTTCCTAGCTCCTACTCTGGGCAGCTAACG

esiRNA cDNA target sequence

ACGGTGTGCCTGTATTGACATCTTTGTTCTGGTTTTGACTTGGCTTGACTTGGTAAGACTAGTCTTTGGAACTTGCCCCACTCCACGTGAGTGGAAGCGTGGCCTGATCACCCACGGTGTGCCTGTATTGGCACTTTTGTTCTGGTTTTGACTTGGCTTGACTTGGTAAGACTAGTCTTTGGAACTTGCCCCACTCCATCTGAGTGGAAACGTGGCCTGATCACCCACAGCATGCCTGCATGGGCACTCTTGTTCTGGTTTTGACTTGACTTGAATTGCTGGATACTTTGGTTTTGGTTTTGACTTGGCTTGAATTTTTTGGTACTCGGATTTTGAATTTCGTGATTTTGGTTTGGTATAAACGGTAAAAGTGTGTGTGCCCTCTTTACCTGTTCTTTGTCTTGTGGTATGTGTGGTGTGAGCGTGGTATT

esiRNA cDNA target sequence

ACTGAGGCCTCTTGCAGAAATCCATATGAGTGGGCCAGCTGACAAGAAAATCTTCCAGCTCTAGTCAAGCCTTCAGAACACCAGCTCTTAGTGCCAGGAACTGCCAGGTAAGAATTCTGACTACACTGAGGCCACCATGCTCTGAGGAAGTCAAGCCAGATGGAAAGATCACAGCAGGAGAACTGGTCAGCAGTCATACTCTTGAACTCATCCCAACCCAGACTCCAGACATGTGAATTGACAAGTCTTCAGATGATTCCAGCACAAAGCCATTGAGTCACCTCCAGCCATTGAGTGTCCCCAGCCGAGACACCAGTCACCATCAAGCAGAGCCAGTATGCCTGTTATGCCTGTCCAAATTCTTGACCCACTAAATTCTATGAAATTAATAAATGGTTGTTTAAGTTACTAAATTTTAAACTTAATTTGTTATGCAGTAATAGGAATTGTTCCTGTCTGTGAGCAAGTCCTCATGGACAATAAACTCAGATCTGTCTGTGGAATCTGGGAGAAAAAGAGACTGTTATCCTTCCCCATGGTACAGCCCCAGAAAGA

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

form

lyophilized powder

form

lyophilized powder

form

lyophilized powder

form

lyophilized powder

NCBI accession no.

XR_113124.1, XR_110877.1, XR_110876.1, XR_113123.1

NCBI accession no.

-

NCBI accession no.

-

NCBI accession no.

-


Storage Class

10 - Combustible liquids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters