Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
| Size/SKU | Availability | Price |
|---|---|---|
20 μg | Please contact Customer Service for Availability | 5 690,00 CZK |
50 μg | Please contact Customer Service for Availability | 10 300,00 CZK |
About This Item
NACRES:
NA.51
UNSPSC Code:
41105324
eCl@ss:
32119207
5 690,00 CZK
Please contact Customer Service for Availability
product line
MISSION®
form
lyophilized, lyophilized powder
esiRNA cDNA target sequence
TCTCTCACAGGGCTGCTACAAAGATTGCATGAGATTTTGCTCAACAAATATTAGCTGTTGGCTCCAGAGTTTAATACCTCTGATCACTACAAGCTTTGAAGGCAAGATAAAATAACTTCAACAGAGTGAATCATCTGTGAACACTTCCTGTGAACGTGTGACTATGATTATGGAACACTCTGTTGCATCAAGATGGCCAGAGATTGTCTCA
shipped in
ambient
storage temp.
−20°C
General description
MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.
For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.
For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.
Legal Information
MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class
11 - Combustible Solids
WGK
WGK 1
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.