Skip to Content
Merck

Skip To

EHU123761

MISSION® esiRNA

targeting human ERLIN2

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
20 μg
Please contact Customer Service for Availability
5 690,00 CZK
50 μg
Please contact Customer Service for Availability
10 300,00 CZK

About This Item

NACRES:
NA.51
UNSPSC Code:
41105324
eCl@ss:
32119207

5 690,00 CZK


Please contact Customer Service for Availability

​
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


description

Powered by Eupheria Biotech

Quality Segment

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

TGGGACTAGTGGTGGTGTGATGATCTACTTTGACAGAATTGAAGTGGTGAACTTCCTGGTCCCGAACGCAGTGTATGATATAGTGAAGAACTATACTGCTGACTATGACAAGGCCCTCATCTTCAACAAGATCCACCACGAACTGAACCAGTTCTGCAGTGTGCACACGCTTCAAGAGGTCTACATTGAGCTGTTTGATCAGATTGATGAAAATCTCAAACTGGCTTTGCAACAGGACCTGACCTCCATGGCCCCTGGGCTGGTCATTCAAGCTGTGCGGGTAACAAAGCCCAACATACCAGAGGCAATCCGCAGAAACTACGAGTTGATGGAAAGTGAGAAGACAAAGCTTCTCATTGCCGCCCAGAAACAGAAGGTGGTGGAAAAGGAAGCAGAGACAGAGCG

Ensembl | human accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

human ... ERLIN2(11160)

General description

MISSION esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
HPA002025EMU014191SAB5701991
description

targeting human ERLIN2

description

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

description

targeting mouse Erlin2

description

-

Ensembl | human accession no.

ENSG00000147475

Ensembl | human accession no.

-

Ensembl | human accession no.

-

Ensembl | human accession no.

-

Gene Information

human ... ERLIN2(11160)

Gene Information

human ... ERLIN2(11160)

Gene Information

mouse ... ERLIN2(244373), Erlin2(244373)

Gene Information

human ... Erlin-2(11160)

esiRNA cDNA target sequence

TGGGACTAGTGGTGGTGTGATGATCTACTTTGACAGAATTGAAGTGGTGAACTTCCTGGTCCCGAACGCAGTGTATGATATAGTGAAGAACTATACTGCTGACTATGACAAGGCCCTCATCTTCAACAAGATCCACCACGAACTGAACCAGTTCTGCAGTGTGCACACGCTTCAAGAGGTCTACATTGAGCTGTTTGATCAGATTGATGAAAATCTCAAACTGGCTTTGCAACAGGACCTGACCTCCATGGCCCCTGGGCTGGTCATTCAAGCTGTGCGGGTAACAAAGCCCAACATACCAGAGGCAATCCGCAGAAACTACGAGTTGATGGAAAGTGAGAAGACAAAGCTTCTCATTGCCGCCCAGAAACAGAAGGTGGTGGAAAAGGAAGCAGAGACAGAGCG

esiRNA cDNA target sequence

-

esiRNA cDNA target sequence

ATCTCATGCTCCCGTTCATCACATCCTATAAGTCTGTACAGACCACTCTCCAAACTGATGAAGTGAAGAACGTACCATGTGGAACCAGTGGTGGTGTGATGATCTACTTTGACAGAATTGAAGTGGTGAACTTCCTGGTCCCAAATGCAGTGTATGATATAGTGAAGAACTATACTGCAGACTATGACAAGGCCCTCATCTTCAACAAGATCCATCATGAGCTTAACCAGTTCTGCAGCGTTCATACTCTTCAGGAAGTCTATATCGAGCTGTTTGATCAAATTGATGAAAACCTCAAGTTGGCTTTGCAGCAGGACCTGACTTCCATGGCCCCTGGGCTGGTTATCCAAGCTGTGCGAGTGACAAAGCCCAATATACCTGAGGCAATCCGCAGGAACTATGAGCTGATGGAAAGCGAGAAGACGAA

esiRNA cDNA target sequence

-

product line

MISSION®

product line

Prestige Antibodies® Powered by Atlas Antibodies

product line

MISSION®

product line

-

NCBI accession no.

NM_001003790

NCBI accession no.

-

NCBI accession no.

NM_153592

NCBI accession no.

-

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

-


Storage Class

10 - Combustible liquids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters