Skip to Content
Merck

Skip To

EMNC010431

MISSION® esiRNA

targeting mouse Mef2c

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
20 μg
Please contact Customer Service for Availability
5 690,00 CZK
50 μg
Please contact Customer Service for Availability
10 300,00 CZK

About This Item

NACRES:
NA.51
UNSPSC Code:
41105324
eCl@ss:
32119207

5 690,00 CZK


Please contact Customer Service for Availability

​
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

TGACTGGGATTTCATGCTCAGCACTGTGCCTTAATTCACAGACTCATGGTTGTTCCTGCATTCAGTAGAGCTTGACTGGGATTTGTCACCTTTAAGAAATCAAACCAGATTACAGAGGCAAGTCACAAAACCAACGCAACAGCGCTCTACACCTTACAAAAGGCAGATTACACAGTTCACATGTTATTCTGATAGAGCAAAGATAAAGGCTGATTGCTAAGGAACGGCAAGACAC

GenBank accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services
MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
EHU113621SAB1401241AMAB90727
description

targeting mouse Mef2c

description

targeting human MEF2C

description

purified immunoglobulin, buffered aqueous solution

description

Prestige Antibodies® Powered by Atlas Antibodies, clone CL0368, purified immunoglobulin, buffered aqueous glycerol solution

esiRNA cDNA target sequence

TGACTGGGATTTCATGCTCAGCACTGTGCCTTAATTCACAGACTCATGGTTGTTCCTGCATTCAGTAGAGCTTGACTGGGATTTGTCACCTTTAAGAAATCAAACCAGATTACAGAGGCAAGTCACAAAACCAACGCAACAGCGCTCTACACCTTACAAAAGGCAGATTACACAGTTCACATGTTATTCTGATAGAGCAAAGATAAAGGCTGATTGCTAAGGAACGGCAAGACAC

esiRNA cDNA target sequence

TGGGTTGATGAAGAAGGCTTATGAGCTGAGCGTGCTGTGTGACTGTGAGATTGCGCTGATCATCTTCAACAGCACCAACAAGCTGTTCCAGTATGCCAGCACCGACATGGACAAAGTGCTTCTCAAGTACACGGAGTACAACGAGCCGCATGAGAGCCGGACAAACTCAGACATCGTGGAGACGTTGAGAAAGAAGGGCCTTAATGGCTGTGACAGCCCAGACCCCGATGCGGACGATTCCGTAGGTCACAGCCCTGAGTCTGAGGACAAGTACAGGAAAATTAACGAAGATATTGATCTAATGATCAGCAGGCAAAGATTGTGTGCTGTTCCACCTCCCAACTTCGAGATGCCAGTCTCCATCCCAGTGTCCAGCCACAACAGTTTGGTGTACAGCAACCCTGTCAGCTCACT

esiRNA cDNA target sequence

-

esiRNA cDNA target sequence

-

product line

MISSION®

product line

MISSION®

product line

-

product line

Prestige Antibodies® Powered by Atlas Antibodies

Gene Information

mouse ... MEF2C(17260), Mef2c(17260)

Gene Information

human ... MEF2C(4208)

Gene Information

human ... MEF2C(4208)

Gene Information

human ... MEF2C(4208)

form

lyophilized powder

form

lyophilized powder

form

buffered aqueous solution

form

buffered aqueous glycerol solution

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

GenBank accession no.

AK036940

GenBank accession no.

-

GenBank accession no.

-

GenBank accession no.

-


Storage Class

12 - Non Combustible Liquids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters