Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
20 μg | Please contact Customer Service for Availability | 5 690,00 CZK |
50 μg | Please contact Customer Service for Availability | 10 300,00 CZK |
About This Item
5 690,00 CZK
product line
MISSION®
form
lyophilized powder
esiRNA cDNA target sequence
CATCTCATGTCCCACCCTCTGAGACCACCCTGCCTCAGAAGAATGCCGGACATTTGGGAGACATGAGAATGCATTTTTTTAAAAAGCCTTCAAGCATTTTGAGAGTAGCTTGTCATAAGCTGACAGTGGGGAGGAATCAGCTCCGTGACTTTCCTGAGTCCCTTCCCATAGGACAAGAGACCTAGCTCTTTGCGTTTGTGCAATAACTACTTGCTAGGAGAGCGGGTAGCACCACCCTCCTCTAGACTACTAACACACAGCACAAGACACATGTCAGCCC
GenBank accession no.
shipped in
ambient
storage temp.
−20°C
Gene Information
mouse ... CUX1(13047)
General description
For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.
Legal Information
1 of 1
This Item | |||
|---|---|---|---|
| description targeting mouse Cux1 | description targeting mouse Cux1 | description targeting human AC093634 | description targeting human ZNF197-AS1 |
| esiRNA cDNA target sequence CATCTCATGTCCCACCCTCTGAGACCACCCTGCCTCAGAAGAATGCCGGACATTTGGGAGACATGAGAATGCATTTTTTTAAAAAGCCTTCAAGCATTTTGAGAGTAGCTTGTCATAAGCTGACAGTGGGGAGGAATCAGCTCCGTGACTTTCCTGAGTCCCTTCCCATAGGACAAGAGACCTAGCTCTTTGCGTTTGTGCAATAACTACTTGCTAGGAGAGCGGGTAGCACCACCCTCCTCTAGACTACTAACACACAGCACAAGACACATGTCAGCCC | esiRNA cDNA target sequence CAAGGGGAGATTGATGCACTGAGTAAAAGAAGCAAAGAAGCAGAGGCAGCCTTCTTGACTGTGTACAAGAGACTAATTGATGTTCCAGATCCGGTACCAGCCCTGGACGTCGGGCAACAGCTGGAAATAAAAGTGCAGCGTCTACACGACATTGAAACAGAGAACCAGAAACTTAGGGAAACACTAGAAGAGTACAACAAGGAGTTTGCTGAAGTGAAAAATCAAGAGGTTACGATAAAAGCACTTAAGGAGAAAATCCGAGAATACGAGCAGACCCTGAAGAGTCAGGCCGAGACCATTGCTCTGGAGAAAGAGCAGAAGCTACAAAATGATTTTGCAGAGAAGGAGAGAAAGCTGCAAGAGACACAGATGTCCACCACCTCAAAACTGGAGGAAGCTGAGCA | esiRNA cDNA target sequence CACTTCTCAACGCACTTGGAGCGGCCCCAGCGCCGCCGCCTGTGTGCGGAGTGCGGAGCCGACTTGGGGAGCGGCGGGGTGCGGGCCGGAGGCAGCCCGGGTACAGGGGTGCGCTTCCCAAGCCCCGTCCGCTGCTCAGAGCATCCCCAAGATTTCCCTCATGGGTCCTGTCCTGGTTGTTTAAGAAGGTTTCCAGAAAACCTCCTGGTTTGAAGAACAAAGATTCGTGGAATTATTGTACCTTGAGAGCTGGAAAGGGCT | esiRNA cDNA target sequence CATGCCCCCAGCTAGTAAGAGGGTTCTATGTGTTGATTCACTGGGAAACCAGAGACTAGAGATACCTGTGAAGACACTTTCAAGAAGTTAGAAGAATTACCCTTGAATGAAGAAAGGAGCTGACTAGAAGTAGTTTATTGAATAACACACAAAGATTGTTTCAAACCTACAAAAGATAAATGTGAGGAAGGGATTTGGGGAATCCTCCTATGTCCTCCAGTCTGGC |
| product line MISSION® | product line MISSION® | product line MISSION® | product line MISSION® |
| Gene Information mouse ... CUX1(13047) | Gene Information mouse ... CUX1(13047), Cux1(13047) | Gene Information - | Gene Information - |
| storage temp. −20°C | storage temp. −20°C | storage temp. −20°C | storage temp. −20°C |
| form lyophilized powder | form lyophilized powder | form lyophilized powder | form lyophilized powder |
| GenBank accession no. | GenBank accession no. - | GenBank accession no. | GenBank accession no. |
Storage Class
12 - Non Combustible Liquids
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
