Skip to Content
Merck

Skip To

EMU024331

MISSION® esiRNA

targeting mouse Cdpf1

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
20 μg
Please contact Customer Service for Availability
5 690,00 CZK
50 μg
Please contact Customer Service for Availability
10 300,00 CZK

About This Item

NACRES:
NA.51
UNSPSC Code:
41105324
eCl@ss:
32119207

5 690,00 CZK


Please contact Customer Service for Availability

​
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


description

Powered by Eupheria Biotech

Quality Segment

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

CCCTGTGTCCAGGAGAACATGAGTGCTTTCCCTCAGGAAATCCAGCAGGATGTAGAGAAGAGAAAGAGCGCCAGCAAGAAGCACTCCAGCCGTCCCTGACTGGCAGGGGCACAGCCCTCCTGCGAAGCATTGGGCTGGGCTCAGCCAGGATTCAGGCTGGGAGCTGTGGACCCGGTCTTCTTTGAACTGGGTGGATACCTCCAGCCTCGGAAGCTGAGGAACTGGCCAGGGCTGAGGACAGACATCAGGGGTGTATCCAGCTGGAGCTCTAGGTCTGGCCTCTGGAGGCCAGGAGAGATGGAGAGGGACCTCCTTTGATTGACAGCTGGTAGACACTCCCTGTAGATGCACACGTGGGAAGACACCTAGCTTCTCTCCAGACTTCCAAACAACACAATGTCATCTGTCTAGGGAATTCAGCCTCCGAGGGCCAGAGGGTAAGCAAGGGTGA

Ensembl | mouse accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
EHU142301HPA018823
description

targeting mouse Cdpf1

description

targeting human CDPF1

description

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

product line

MISSION®

product line

MISSION®

product line

Prestige Antibodies® Powered by Atlas Antibodies

Ensembl | mouse accession no.

ENSMUSG00000064284

Ensembl | mouse accession no.

-

Ensembl | mouse accession no.

-

esiRNA cDNA target sequence

CCCTGTGTCCAGGAGAACATGAGTGCTTTCCCTCAGGAAATCCAGCAGGATGTAGAGAAGAGAAAGAGCGCCAGCAAGAAGCACTCCAGCCGTCCCTGACTGGCAGGGGCACAGCCCTCCTGCGAAGCATTGGGCTGGGCTCAGCCAGGATTCAGGCTGGGAGCTGTGGACCCGGTCTTCTTTGAACTGGGTGGATACCTCCAGCCTCGGAAGCTGAGGAACTGGCCAGGGCTGAGGACAGACATCAGGGGTGTATCCAGCTGGAGCTCTAGGTCTGGCCTCTGGAGGCCAGGAGAGATGGAGAGGGACCTCCTTTGATTGACAGCTGGTAGACACTCCCTGTAGATGCACACGTGGGAAGACACCTAGCTTCTCTCCAGACTTCCAAACAACACAATGTCATCTGTCTAGGGAATTCAGCCTCCGAGGGCCAGAGGGTAAGCAAGGGTGA

esiRNA cDNA target sequence

CCCAAAGAACCAGCTCATGTTTGGAAAAGGAATCTTCCACTGAAACATTTGGATACCCGACGTTCTCCCTGGACACTTGGCCCGGCAATAGTTGGGGTCTCCTGGGAAGCTGAGTGCTGGCTTCTAGCCTCTCCAGTTCCCTCAGACTCTGTCTTCAGCTCGGGCCCCTCCGTTCTGTCTCCCTCGACTCCAAGTCAGGAATACCGTGGGCTTCCTGCCCTCACCCCACACCTCAAGACCATCCCACTGCTTGCCTGCTTGCCCCCTGCCATCTCGCACTCAGCCCTGGGTGCCTGTGAAGCCTGGCCCCACTGCCTGCCGGGGGTTCCTGCAGCACCCCCCTACTCCAGGCTCCCCATCTCCCTGAAGCTTCCAGGGTAGCAGCTGGTGGGACACTGATCTGTGAGCTCTGGAATGTCTGCTTAGAATTCTGAGAGGCAGGAACTGCTC

esiRNA cDNA target sequence

-

Gene Information

mouse ... CDPF1(72355), 2210021J22Rik(72355)

Gene Information

human ... CDPF1(150383), C22orf40(150383)

Gene Information

human ... C22orf40(150383)

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

NCBI accession no.

NM_197998

NCBI accession no.

NM_207327

NCBI accession no.

-


Storage Class

10 - Combustible liquids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters