Skip to Content
Merck

Skip To

EMU165151

MISSION® esiRNA

targeting mouse Zfp784

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
20 μg
Please contact Customer Service for Availability
5 690,00 CZK
50 μg
Please contact Customer Service for Availability
10 300,00 CZK

About This Item

NACRES:
NA.51
UNSPSC Code:
41105324
eCl@ss:
32119207

5 690,00 CZK


Please contact Customer Service for Availability

​
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


description

Powered by Eupheria Biotech

Quality Segment

product line

MISSION®

form

lyophilized powder

esiRNA cDNA target sequence

TGGGCTCTCTTTAACCCACTAAACTTAGTTGCAAGCAACTGAGAGCACACACTATAAGCAGGCAATCACTTGGTATCCCTTTAAGCCTTAGGCTCAGAACCTGTCTATAAAGAAATTTCCCATCTCTCTATTCTGCACTCTGAAGTAGGCATCTTCCTGCATAGCCCTGGGCAATTCTGAGGCAGGGTTAGCTAGTGACCACCTTGGTGGGCCAGTACAGGGTGTAAAGAAAGAAATGACACTGTGGTAAGGGGCTAAAAGGACTTGGGAGTTAGCGTACAGGGTAAGAGTGGGGACTTCCAGTCTGTTCCTATTAAAAGACTTTCTGTAAGTCTTCTCTGTCAGTCTTGTATTCCAATTCTCTGGGTAGTTATCCATTATCTGGAATGGTGAGTGACCAGGCTTGGGCTGTTTGGAGATGTCTATTTGGTTGTGTTGTAAGAAGTTTTGTGCAATGAGGTGATGTAGGAGGTTGGTGTACTTACTCAGAAGGGTCCCTAATTTTCTCATG

Ensembl | mouse accession no.

NCBI accession no.

shipped in

ambient

storage temp.

−20°C

Gene Information

General description

MISSION® esiRNA are endoribonuclease prepared siRNA. They are a heterogeneous mixture of siRNA that all target the same mRNA sequence. These multiple silencing triggers lead to highly-specific and effective gene silencing.

For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.

Legal Information

MISSION is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
EMU173901
description

targeting mouse Zfp784

description

targeting mouse Zfp865

Ensembl | mouse accession no.

ENSMUSG00000043290

Ensembl | mouse accession no.

ENSMUSG00000074405

product line

MISSION®

product line

MISSION®

esiRNA cDNA target sequence

TGGGCTCTCTTTAACCCACTAAACTTAGTTGCAAGCAACTGAGAGCACACACTATAAGCAGGCAATCACTTGGTATCCCTTTAAGCCTTAGGCTCAGAACCTGTCTATAAAGAAATTTCCCATCTCTCTATTCTGCACTCTGAAGTAGGCATCTTCCTGCATAGCCCTGGGCAATTCTGAGGCAGGGTTAGCTAGTGACCACCTTGGTGGGCCAGTACAGGGTGTAAAGAAAGAAATGACACTGTGGTAAGGGGCTAAAAGGACTTGGGAGTTAGCGTACAGGGTAAGAGTGGGGACTTCCAGTCTGTTCCTATTAAAAGACTTTCTGTAAGTCTTCTCTGTCAGTCTTGTATTCCAATTCTCTGGGTAGTTATCCATTATCTGGAATGGTGAGTGACCAGGCTTGGGCTGTTTGGAGATGTCTATTTGGTTGTGTTGTAAGAAGTTTTGTGCAATGAGGTGATGTAGGAGGTTGGTGTACTTACTCAGAAGGGTCCCTAATTTTCTCATG

esiRNA cDNA target sequence

TCCTCCACCCCAATATGACTACCCACCCCAGTCCAGTTTCAAACCCAAGGCGGAGGCTCCTTCCTCATCTTCATCATCATCTTCATCATCATCTTCCTCATCTTCGTCTTCCTCATCCTCTTCCCAAGCCAAGAAGATGGATCCGCCTTTGCCACCCACCTTCGGGGCACCTCCCCCTCCACTCTTTGATGCTGCCTTCCCTGCTCCGCAATGGGGAATAGTCGACCTCTCTGGACACCAGCACCTGTTCGGGAACCTGAAACGTGGTGGGCCCACCCCTGGGCCTGGAGTAGCATCAGGACTGGGCACTCCCACTGGGACCCCAGGGCCACTTACCACACCCTCGCAGACCCCGCCTGGGCCCGCAGTAGGGGCAGCCTGCGACCCAACCAAGGACGACAAGGGCTATTTCCGAAGGCTGAAGTACCTGATGGAGCGACGCTTCCCGTGTGGTGTTTGCCAGAAGTCCTTCAAACAGTCCTCACATCTGGTCCAGCACATGTTAGTGCACTCCG

Gene Information

mouse ... ZFP784(319748), Zfp784(654801)

Gene Information

mouse ... ZFP865(319748), Zfp865(319748)

form

lyophilized powder

form

lyophilized powder

storage temp.

−20°C

storage temp.

−20°C


Storage Class

10 - Combustible liquids

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters