Select a Size
| Size/SKU | Availability | Price |
|---|---|---|
20 μg | Please contact Customer Service for Availability | 5 690,00 CZK |
50 μg | Please contact Customer Service for Availability | 10 300,00 CZK |
About This Item
5 690,00 CZK
description
Powered by Eupheria Biotech
Quality Segment
product line
MISSION®
form
lyophilized powder
esiRNA cDNA target sequence
TGAGCCACGTGAATCTGTAACATTTGAGGATGTGTCTGTGGATTTCTCCAGAGGAGAGTGGAAGCTACTGGAACCATCTCAAAGAGAGCTATATAAGGAAGTGCTGCTGGAAAACTTAGGGACCCTAGAGTTTTTGGGCTCTCCGGTTTCCAAACTTGACTTGATTTCCCATCTGAAGTGGGTTAAATTGCCAAGGGTACTGGAAAAAGAAATCTTAAAAGACCCCAGACCAGGAGAGTTGGTAAAAGAGAGTGAGTCTAGACGTGAATTGGATGCTTTTATGGAAGGCTTGACTTTAGAGAAAACAGTGGAGTACTGCTTCAGGGATGGTGGTTATGGCTTGAAGACAGAATTCCAGAGGCGACATGGCAAACCCAAGAAAGATCATGGCAAGCGGAGTAGCCATGAGAATAGAGAATCTGAGACAGAAGGGACCTCCTCTGGGAAGAGAGTCAAGCAGACTTCTGACACACTCAAACATCGGAAAGCCTCCTTATCTAAGATGTCTCA
Ensembl | mouse accession no.
NCBI accession no.
shipped in
ambient
storage temp.
−20°C
Gene Information
mouse ... ZKSCAN16(100041581), Zkscan16(100041581)
General description
For additional details as well as to view all available esiRNA options, please visit SigmaAldrich.com/esiRNA.
Legal Information
1 of 1
This Item | |
|---|---|
| description targeting mouse Zkscan16 | description Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution |
| Ensembl | mouse accession no. | Ensembl | mouse accession no. - |
| Gene Information mouse ... ZKSCAN16(100041581), Zkscan16(100041581) | Gene Information human ... ZNF483(158399) |
| esiRNA cDNA target sequence TGAGCCACGTGAATCTGTAACATTTGAGGATGTGTCTGTGGATTTCTCCAGAGGAGAGTGGAAGCTACTGGAACCATCTCAAAGAGAGCTATATAAGGAAGTGCTGCTGGAAAACTTAGGGACCCTAGAGTTTTTGGGCTCTCCGGTTTCCAAACTTGACTTGATTTCCCATCTGAAGTGGGTTAAATTGCCAAGGGTACTGGAAAAAGAAATCTTAAAAGACCCCAGACCAGGAGAGTTGGTAAAAGAGAGTGAGTCTAGACGTGAATTGGATGCTTTTATGGAAGGCTTGACTTTAGAGAAAACAGTGGAGTACTGCTTCAGGGATGGTGGTTATGGCTTGAAGACAGAATTCCAGAGGCGACATGGCAAACCCAAGAAAGATCATGGCAAGCGGAGTAGCCATGAGAATAGAGAATCTGAGACAGAAGGGACCTCCTCTGGGAAGAGAGTCAAGCAGACTTCTGACACACTCAAACATCGGAAAGCCTCCTTATCTAAGATGTCTCA | esiRNA cDNA target sequence - |
| product line MISSION® | product line Prestige Antibodies® Powered by Atlas Antibodies |
| storage temp. −20°C | storage temp. −20°C |
| NCBI accession no. | NCBI accession no. - |
Storage Class
10 - Combustible liquids
Flash Point (°F)
Not applicable
Flash Point (°C)
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.

