material
stainless steel
feature
to hold 12 x 53 mm/ 2 in boxes (format 6 x 2)
for use with CryoCube® F101h
packaging
pack of 1 ea
manufacturer/tradename
Eppendorf® 6001092210
D × W × H
-307 mm -12.1 in. -140 mm -5.5 in. -337 mm -13.3 in.
General description
Stainless steel, drawer style, side access racks that give easy access to samples without removing most of the storage boxes from the cold interior. Each rack has 6 drawers and each drawer can hold 2 boxes with dimensions 136mm square and 53mm high. The side access rack must be completely removed for sample access.
Application
cryostorage
Features and Benefits
Also suitable for use as drawer rack
Legal Information
CryoCube is a registered trademark of Eppendorf AG
Eppendorf is a registered trademark of Eppendorf AG
Choose from one of the most recent versions:
Already Own This Product?