Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C4H10N3O5P · 2C4H11NO3
CAS Number:
Molecular Weight:
453.38
UNSPSC Code:
12352200
PubChem Substance ID:
MDL number:
assay
~98%
storage temp.
−20°C
SMILES string
NC(CO)(CO)CO.NC(CO)(CO)CO.CN(CC(O)=O)C(=N)NP(O)(O)=O
InChI
1S/C4H10N3O5P.2C4H11NO3/c1-7(2-3(8)9)4(5)6-13(10,11)12;2*5-4(1-6,2-7)3-8/h2H2,1H3,(H,8,9)(H4,5,6,10,11,12);2*6-8H,1-3,5H2
InChI key
UPPBKCOGWJTQMJ-UHFFFAOYSA-N
Application
For use in systems where alkali metal ions are undesirable.
Biochem/physiol Actions
High-energy phosphate reservoir in vertebrate and some invertebrate muscle; provides phosphate for ADP-ATP conversion
High-energy phosphate reservoir in vertebrate and some invertebrate muscle; provides phosphate for ADP-ATP conversion. As a non-oxidative reservoir of high-energy phosphate, phosphocreatine protects against anoxic and ischemic damage to brain neurons.
Storage Class
11 - Combustible Solids
wgk
WGK 3
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, type N95 (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Sofía Muñoz et al.
Molecular cell, 74(4), 664-673 (2019-03-30)
Cohesin is a conserved, ring-shaped protein complex that topologically embraces DNA. Its central role in genome organization includes functions in sister chromatid cohesion, DNA repair, and transcriptional regulation. Cohesin loading onto chromosomes requires the Scc2-Scc4 cohesin loader, whose presence on
Lou Beaulieu-Laroche et al.
Neuron, 103(2), 235-241 (2019-06-11)
Dendritic integration can expand the information-processing capabilities of neurons. However, the recruitment of active dendritic processing in vivo and its relationship to somatic activity remain poorly understood. Here, we use two-photon GCaMP6f imaging to simultaneously monitor dendritic and somatic compartments in