Skip to Content
Merck

SAB4300719

Anti-SFTPA1 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

PSAP, PSPA, SP-A, SPA, surfactant protein A1

Sign In to View Organizational & Contract Pricing.

Select a Size

100 μL

CZK 9,280.00

CZK 9,280.00


Please contact Customer Service for Availability


About This Item

NACRES:
NA.41
UNSPSC Code:
12352203

Skip To

Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist

Product Name

Anti-SFTPA1 antibody produced in rabbit, affinity isolated antibody

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

28 kDa

species reactivity

human

concentration

1.3 mg/mL

technique(s)

western blot: 1:200-1:1000 (Cell Lysate)

isotype

IgG

accession no.

NP_001087239.2

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Quality Level

Gene Information

human ... SFTPA1(653509)

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
HPA042638HPA045752AV09053
biological source

rabbit

biological source

rabbit

biological source

rabbit

biological source

rabbit

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

affinity isolated antibody

antibody form

IgG fraction of antiserum

species reactivity

human

species reactivity

human

species reactivity

human

species reactivity

mouse, human, guinea pig, horse, dog, rat

mol wt

28 kDa

mol wt

-

mol wt

-

mol wt

35 kDa

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

Biochem/physiol Actions

The antibody detects endogenous level of total SFTPA1 protein.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Features and Benefits

Evaluate our antibodies with complete peace of mind. If the antibody does not perform in your application, we will issue a full credit or replacement antibody. Learn more.

Immunogen

Synthetic peptide corresponding to a region derived from 223-236 amino acids of human surfactant protein A1

Physical form

Supplied at 1.3mg/mL in phosphate buffered saline (without Mg2+ and Ca2+), pH 7.3, 0.05% sodium azide and 50% glycerol.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zhong-Yi Sun et al.
Molecular medicine reports, 19(6), 4980-4988 (2019-05-07)
Deficiency of surfactant proteins (SPs) is the main cause of respiratory distress syndrome (RDS) and chronic lung diseases. Our previous study demonstrated that miR‑431 was differentially expressed between infants with RDS and infants without RDS using microarray analysis. However, the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service