Skip to Content
Merck

Skip To

SML2275

TH-257

≥98% (HPLC)

Synonym(s):

N-Benzyl-N-butyl-4-(N-phenylsulfamoyl)benzamide, N-Butyl-4-[(phenylamino)sulfonyl]-N-(phenylmethyl)benzamide

Sign In to View Organizational & Contract Pricing.

About This Item

Empirical Formula (Hill Notation):
C24H26N2O3S
CAS Number:
Molecular Weight:
422.54
UNSPSC Code:
12352200
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
powder


assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(C1=CC=C(C=C1)S(NC2=CC=CC=C2)(=O)=O)N(CC3=CC=CC=C3)CCCC

InChI

1S/C24H26N2O3S/c1-2-3-18-26(19-20-10-6-4-7-11-20)24(27)21-14-16-23(17-15-21)30(28,29)25-22-12-8-5-9-13-22/h4-17,25H,2-3,18-19H2,1H3

InChI key

VNCIWNGCMAKKEO-UHFFFAOYSA-N

Biochem/physiol Actions

Potent, type III allosteric LIM-kinase (LIMK1 & LIMK2) inhibitor that targets LIMK1/2 DFG-out conformation in a non-ATP-competitive manner.
TH-257 is a potent, type III allosteric LIM-kinase (LIMK1 & LIMK2) inhibitor (LIMK1/2 IC50 in nM = 48/29 by cell-free kinase assay and 250/150 by live cell NanoBRET assay using respective transfectants) that targets LIMK1/2 DFG-out conformation in a non-ATP-competitive manner and exhibits no significant off-target activity by KINOMEscan assay at 1 μM. TH-263 is a structure analog and the recommended negative control for TH-257. For characterization details of TH-257, please visit the TH-257 probe summary on the Structural Genomics Consortium (SGC) website.

TH-263 is the negative control for the active probe, TH-257. TH-263 is available from Sigma. To learn more about and purchase TH-263, click here.

To learn about other SGC chemical probes, visit sigma.com/sgc

Features and Benefits

TH-257 is a chemical probe available through a partnership with the Structural Genomics Consortium (SGC). To learn more and view other SGC epigenetic probes, visit sigma.com/SGC.


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library