Merck
  • Targeting Breast Cancer Stem Cell State Equilibrium through Modulation of Redox Signaling.
Targeting Breast Cancer Stem Cell State Equilibrium through Modulation of Redox Signaling.
Cell metabolism (2018-07-05)
Ming Luo, Li Shang, Michael D Brooks, Evelyn Jiagge, Yongyou Zhu, Johanna M Buschhaus, Sarah Conley, Melissa A Fath, April Davis, Elizabeth Gheordunescu, Yongfang Wang, Ramdane Harouaka, Ann Lozier, Daniel Triner, Sean McDermott, Sofia D Merajver, Gary D Luker, Douglas R Spitz, Max S Wicha
ABSTRACT
Although breast cancer stem cells (BCSCs) display plasticity transitioning between quiescent mesenchymal-like (M) and proliferative epithelial-like (E) states, how this plasticity is regulated by metabolic or oxidative stress remains poorly understood. Here, we show that M- and E-BCSCs rely on distinct metabolic pathways and display markedly different sensitivities to inhibitors of glycolysis and redox metabolism. Metabolic or oxidative stress generated by 2DG, H2O2, or hypoxia promotes the transition of ROSlo M-BCSCs to a ROShi E-state. This transition is reversed by N-acetylcysteine and mediated by activation of the AMPK-HIF1α axis. Moreover, E-BCSCs exhibit robust NRF2-mediated antioxidant responses, rendering them vulnerable to ROS-induced differentiation and cytotoxicity following suppression of NRF2 or downstream thioredoxin (TXN) and glutathione (GSH) antioxidant pathways. Co-inhibition of glycolysis and TXN and GSH pathways suppresses tumor growth, tumor-initiating potential, and metastasis by eliminating both M- and E-BCSCs. Exploiting metabolic vulnerabilities of distinct BCSC states provides a novel therapeutic approach targeting this critical tumor cell population.
Sigma-Aldrich
2-Deoxy-D-glucose, ≥98% (GC), crystalline
Sigma-Aldrich
Diethylenetriaminepentaacetic acid, ≥99% (titration)
Sigma-Aldrich
Amyloid Protein Non-Aβ Component, ≥80% (HPLC)
Sigma-Aldrich
L-Buthionine-sulfoximine, ≥97% (TLC)
Sigma-Aldrich
5,5′-Dithiobis(2-nitrobenzoic acid), ≥98%, BioReagent, suitable for determination of sulfhydryl groups
Sigma-Aldrich
AICAR, ≥98% (HPLC), powder
Sigma-Aldrich
MitoPQ, ≥90% (HPLC)
Sigma-Aldrich
Superoxide dismutase–polyethylene glycol from bovine erythrocytes, lyophilized powder, ≥1,350 units/mg protein
Sigma-Aldrich
Catalase−polyethylene glycol, lyophilized powder, ~40,000 units/mg protein
Sigma-Aldrich
Thioredoxin Reductase Assay Kit, 1 kit sufficient for 100 assays (1 mL)
Sigma-Aldrich
Anti-β-Actin−Peroxidase antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Sigma-Aldrich
2-Vinylpyridine, 97%
Sigma-Aldrich
N-Acetyl-L-cysteine, Sigma Grade, ≥99% (TLC), powder
Roche
cOmplete, Mini, EDTA-free Protease Inhibitor Cocktail, Tablets provided in EASYpacks
Sigma-Aldrich
MISSION® esiRNA, targeting human GABPA
Sigma-Aldrich
γ-Secretase Inhibitor I, The γ-Secretase Inhibitor I controls the biological activity of γ-Secretase. This small molecule/inhibitor is primarily used for Neuroscience applications.