Merck
  • β1 Integrins Are Required To Mediate NK Cell Killing of Cryptococcus neoformans.
β1 Integrins Are Required To Mediate NK Cell Killing of Cryptococcus neoformans.
Journal of immunology (Baltimore, Md. : 1950) (2018-09-12)
Richard F Xiang, ShuShun Li, Henry Ogbomo, Danuta Stack, Christopher H Mody
ABSTRACT
Cryptococcus neoformans is a fungal pathogen that causes fatal meningitis and pneumonia. During host defense to Cryptococcus, NK cells directly recognize and kill C. neoformans using cytolytic degranulation analogous to killing of tumor cells. This fungal killing requires independent activation of Src family kinase (SFK) and Rac1-mediated pathways. Recognition of C. neoformans requires the natural cytotoxicity receptor, NKp30; however, it is not known whether NKp30 activates both signal transduction pathways or whether a second receptor is involved in activation of one of the pathways. We used primary human NK cells and a human NK cell line and found that NKp30 activates SFK → PI3K but not Rac1 cytotoxic signaling, which led to a search for the receptor leading to Rac1 activation. We found that NK cells require integrin-linked kinase (ILK) to activate Rac1 for effective fungal killing. This observation led to our identification of β1 integrin as an essential anticryptococcal receptor. These findings demonstrate that multiple receptors, including β1 integrins and NKp30 and their proximal signaling pathways, are required for recognition of Cryptococcus, which activates a central cytolytic antimicrobial pathway leading to fungal killing.
Sigma-Aldrich
Anti-phospho-ILK (Ser246) Antibody, from rabbit, purified by affinity chromatography
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Methyl-β-cyclodextrin, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Rac1/cdc42 Activation Magnetic Beads Pulldown Assay, The Rac1/cdc42 Activation Magnetic Beads Pulldown Assay provides an effective method for detecting Rac & Cdc42 activity in cell lysates with higher yield & easier process utilizing magenetic bead properties.
Sigma-Aldrich
ILK inhibitor, Cpd 22, The ILK inhibitor, Cpd 22 controls the biological activity of ILK. This small molecule/inhibitor is primarily used for Phosphorylation & Dephosphorylation applications.