Merck
  • Overexpression of cell surface cytokeratin 8 in multidrug-resistant MCF-7/MX cells enhances cell adhesion to the extracellular matrix.
Overexpression of cell surface cytokeratin 8 in multidrug-resistant MCF-7/MX cells enhances cell adhesion to the extracellular matrix.
Neoplasia (New York, N.Y.) (2008-10-28)
Fang Liu, Zhong Chen, Jinhong Wang, Xiaofeng Shao, Ziyou Cui, Chunzheng Yang, Zhenping Zhu, Dongsheng Xiong
ABSTRACT
Accumulating evidence suggests that multiple complex mechanisms may be involved, simultaneously or complementarily, in the emergence and development of multidrug resistance (MDR) in various cancers. Cell adhesion-mediated MDR is one such mechanism. In the present study, we initially observed increased cell adhesion to extracellular matrix proteins by the MDR human breast tumor cell line MCF-7/MX compared to its parental cells. We then used a strategy that combined antibody-based screening technique and mass spectrometry-based proteomics to identify membrane proteins that contribute to the enhanced adhesion of MCF-7/MX cells. Using MCF-7/MX cells as immunogen, we isolated a mouse monoclonal antibody, 9C6, that preferentially reacts with MCF-7/MX cells over the parental MCF-7 cells. The molecular target of 9C6 was identified as cytokeratin 8 (CK8), which was found to be overexpressed on the cell surface of MCF-7/MX cells. We further observed that down-regulation of cell surface levels of CK8 through siRNA transfection significantly inhibited MCF-7/MX cell adhesion to fibronectin and vitronectin. In addition, anti-CK8 siRNA partially reversed the MDR phenotype of MCF-7/MX cells. Taken together, our results suggest that alterations in the expression level and cellular localization of CK8 may play a significant role in enhancing the cellular adhesion of MDR MCF-7/MX cells.
Product Description
Millipore
Protein A–Agarose, saline suspension
Millipore
Protein A–Agarose, lyophilized powder
Millipore
Protein A–Agarose, lyophilized powder
Sigma-Aldrich
Fibronectin rat plasma, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Sulforhodamine B, Dye content 75 %
Sigma-Aldrich
DAPI, dilactate, ≥98% (HPLC)
Sigma-Aldrich
Fibronectin human plasma, lyophilized powder, BioReagent, suitable for cell culture
Millipore
Protein G–Agarose, lyophilized powder, Contains lactose stabilizers that must be removed prior to use.
Sigma-Aldrich
FIBRONECTIN FROM HUMAN PLASMA, liquid, 0.1% (Solution), BioReagent, suitable for cell culture
Sigma-Aldrich
Fibronectin bovine plasma, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Fibronectin bovine plasma, solution, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Fibronectin human plasma, lyophilized powder, BioReagent, suitable for cell culture, ≥95% (SDS-PAGE)