Merck
  • Novel targets for Huntington's disease in an mTOR-independent autophagy pathway.
Novel targets for Huntington's disease in an mTOR-independent autophagy pathway.
Nature chemical biology (2008-04-09)
Andrea Williams, Sovan Sarkar, Paul Cuddon, Evangelia K Ttofi, Shinji Saiki, Farah H Siddiqi, Luca Jahreiss, Angeleen Fleming, Dean Pask, Paul Goldsmith, Cahir J O'Kane, Rodrigo Andres Floto, David C Rubinsztein
ABSTRACT
Autophagy is a major clearance route for intracellular aggregate-prone proteins causing diseases such as Huntington's disease. Autophagy induction with the mTOR inhibitor rapamycin accelerates clearance of these toxic substrates. As rapamycin has nontrivial side effects, we screened FDA-approved drugs to identify new autophagy-inducing pathways. We found that L-type Ca2+ channel antagonists, the K+ATP channel opener minoxidil, and the G(i) signaling activator clonidine induce autophagy. These drugs revealed a cyclical mTOR-independent pathway regulating autophagy, in which cAMP regulates IP3 levels, influencing calpain activity, which completes the cycle by cleaving and activating G(s)alpha, which regulates cAMP levels. This pathway has numerous potential points where autophagy can be induced, and we provide proof of principle for therapeutic relevance in Huntington's disease using mammalian cell, fly and zebrafish models. Our data also suggest that insults that elevate intracytosolic Ca2+ (like excitotoxicity) inhibit autophagy, thus retarding clearance of aggregate-prone proteins.
Sigma-Aldrich
Calpain Inhibitor II, powder
Sigma-Aldrich
2-Propylpentanoic acid
Sigma-Aldrich
Thapsigargin, ≥98% (HPLC), solid film
Sigma-Aldrich
Forskolin, For use in molecular biology applications
Sigma-Aldrich
Forskolin, from Coleus forskohlii, ≥98% (HPLC), powder
Sigma-Aldrich
Nitrendipine, >95%, powder