Merck
  • A practical deuterium-free NMR method for the rapid determination of 1-octanol/water partition coefficients of pharmaceutical agents.
A practical deuterium-free NMR method for the rapid determination of 1-octanol/water partition coefficients of pharmaceutical agents.
Bioorganic & medicinal chemistry letters (2010-09-25)
Huaping Mo, Kathryn M Balko, David A Colby
ABSTRACT
A simple and rapid NMR method is described to determine the logP of pharmaceutical agents. This method is highly versatile and efficient, because it does not require the use of deuterated solvents or the addition of any internal/external standards to the sample. We demonstrate that logP can be accurately measured using NMR for pharmaceutical agents with known logP values. Our proposed method is made possible by the combination of state-of-the-art NMR techniques including the solvent concentration reference and robust solvent suppressions.
Sigma-Aldrich
Erythromycin, meets USP testing specifications
Sigma-Aldrich
Erythromycin, tested according to Ph. Eur.
Sigma-Aldrich
Phenacetin, ≥98.0% (HPLC)
Sigma-Aldrich
Erythromycin, BioReagent, suitable for cell culture
Sigma-Aldrich
Acetylsalicylic acid, analytical standard
Sigma-Aldrich
Acetylsalicylic acid, ≥99.0%
Sigma-Aldrich
Lidocaine, powder
Supelco
Nitrofurantoin, VETRANAL®, analytical standard
Sigma-Aldrich
Nitrofurantoin, 98.0-102.0% (EP, UV), meets EP testing specifications
Sigma-Aldrich
Erythromycin, potency: ≥850 μg per mg
Sigma-Aldrich
Lidocaine, analytical standard
Sigma-Aldrich
(S)-(+)-6-Methoxy-α-methyl-2-naphthaleneacetic acid, 98%
Sigma-Aldrich
Naproxen, meets USP testing specifications
Supelco
Naproxen, VETRANAL®, analytical standard
Sigma-Aldrich
Aspirin, meets USP testing specifications
Sigma-Aldrich
Acetaminophen, meets USP testing specifications, 98.0-102.0%, powder
Sigma-Aldrich
Acetaminophen, BioXtra, ≥99.0%
Sigma-Aldrich
Acetaminophen, analytical standard