Merck
Nutrient requirements of suspension cultures of soybean root cells.
Experimental cell research (1968-04-01)
O L Gamborg, R A Miller, K Ojima
Sigma-Aldrich
Gamborg′s Vitamin Solution, 1000 ×, liquid, suitable for plant cell culture
Sigma-Aldrich
Gamborg′s B-5 Basal Medium with Minimal Organics, fine powder, suitable for plant cell culture
Sigma-Aldrich
Murashige and Skoog Basal Medium, powder, suitable for plant cell culture, with Gamborg′s vitamins
Sigma-Aldrich
Gamborg′s B-5 Basal Salt Mixture, fine powder, suitable for plant cell culture