Merck
  • Global identification of multiple OsGH9 family members and their involvement in cellulose crystallinity modification in rice.
Global identification of multiple OsGH9 family members and their involvement in cellulose crystallinity modification in rice.
PloS one (2013-01-12)
Guosheng Xie, Bo Yang, Zhengdan Xu, Fengcheng Li, Kai Guo, Mingliang Zhang, Lingqiang Wang, Weihua Zou, Yanting Wang, Liangcai Peng
ABSTRACT
Plant glycoside hydrolase family 9 (GH9) comprises typical endo-β-1,4-glucanase (EGases, EC3.2.1.4). Although GH9A (KORRIGAN) family genes have been reported to be involved in cellulose biosynthesis in plants, much remains unknown about other GH9 subclasses. In this study, we observed a global gene co-expression profiling and conducted a correlation analysis between OsGH9 and OsCESA among 66 tissues covering most periods of life cycles in 2 rice varieties. Our results showed that OsGH9A3 and B5 possessed an extremely high co-expression with OsCESA1, 3, and 8 typical for cellulose biosynthesis in rice. Using two distinct rice non-GH9 mutants and wild type, we performed integrative analysis of gene expression level by qRT-PCR, cellulase activities in situ and in vitro, and lignocellulose crystallinity index (CrI) in four internodes of stem tissues. For the first time, OsGH9B1, 3, and 16 were characterized with the potential role in lignocellulose crystallinity alteration in rice, whereas OsGH9A3 and B5 were suggested for cellulose biosynthesis. In addition, phylogenetic analysis and gene co-expression comparison revealed GH9 function similarity in Arabidopsis and rice. Hence, the data can provide insights into GH9 function in plants and offer the potential strategy for genetic manipulation of plant cell wall using the five aforementioned novel OsGH9 genes.
Sigma-Aldrich
Cellulase from Trichoderma reesei ATCC 26921, lyophilized powder, ≥1 unit/mg solid
Sigma-Aldrich
Cellulase from Trichoderma reesei, aqueous solution, ≥700 units/g
Sigma-Aldrich
Cellulase from Trichoderma sp., BioReagent, suitable for plant cell culture, 3-10 units/mg solid
Sigma-Aldrich
Cellulase from Trichoderma sp., powder, ≥5,000 units/g solid
Sigma-Aldrich
Cellulase from Aspergillus sp., aqueous solution
Sigma-Aldrich
Cellulase from Aspergillus niger, powder, off-white, ~0.8 U/mg
Sigma-Aldrich
Cellulase from Aspergillus niger, powder, ≥0.3 units/mg solid