Merck
  • Effective modifications for improved homologous recombination and high-efficiency generation of recombinant adenovirus-based vectors.
Effective modifications for improved homologous recombination and high-efficiency generation of recombinant adenovirus-based vectors.
Journal of virological methods (2008-08-30)
Chengxiang Wu, Vivek R Nerurkar, Richard Yanagihara, Yuanan Lu
ABSTRACT
Generation of adenovirus-based vectors through homologous recombination within Escherichia coli cells is one of the most efficient strategies. A common challenge associated with this method is the formation of colonies containing self-ligated shuttle plasmid. To improve homologous recombination, a new pAdEasy-1-bearing competent cell line was constructed so that it no longer requires co-transformation with two plasmids and can generate more recombinant colonies (ninefold). New and efficient approaches were also tested to block shuttle plasmid self-ligation by a combined treatment of the plasmid with Taq DNA polymerase and calf intestine phosphatase (CIP) or blocking the formation of self-ligated plasmid-containing colonies by subcloning a suicide gene, ccdB, into the plasmid construct. Present experimental data show that these modifications are effective in eliminating self-ligated plasmid-containing colony background and offer greater simplicity, faster experimental progress, and higher efficiency in performing homologous recombination within E. coli cells, which could facilitate the production of high-titer infectious viral particles.
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
D-Glutamine, ≥98% (HPLC)
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Glycerol, Molecular Biology, ≥99.0%
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
SAFC
L-Glutamine
Sigma-Aldrich
Glycerol, tested according to Ph. Eur., anhydrous
Supelco
L-Glutamine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Glycerol, BioUltra, Molecular Biology, anhydrous, ≥99.5% (GC)
Supelco
L-Glutamine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Glycerol, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Glycerol, ACS reagent, ≥99.5%