Merck
  • Short-chain fatty acids suppress lipopolysaccharide-induced production of nitric oxide and proinflammatory cytokines through inhibition of NF-κB pathway in RAW264.7 cells.
Short-chain fatty acids suppress lipopolysaccharide-induced production of nitric oxide and proinflammatory cytokines through inhibition of NF-κB pathway in RAW264.7 cells.
Inflammation (2012-06-07)
Tengfei Liu, Jing Li, Yuxin Liu, Nan Xiao, Haitao Suo, Kun Xie, Chunliu Yang, Chen Wu
ABSTRACT
Short-chain fatty acids (SCFAs) produced by the colonic bacterial fermentation of dietary fiber contribute a significant proportion of daily energy requirement. Furthermore, these compounds are modulators of macrophage function and potential targets for the development of new drugs. The aims of this study were to evaluate the effects of three types of SCFAs (sodium acetate (NaAc), sodium propionate (NaP), and sodium butyrate (NaB)) on the production of NO and inducible nitric oxide synthase (iNOS) and proinflammatory and antiinflammatory cytokines (tumor necrosis factor-α (TNF-α) and interleukin (IL-1, IL-6, and IL-10)) and to observe the effect of NaAc on inhibiting lipopolysaccharide (LPS)-induced NF-κB activation in LPS-stimulated RAW264.7 cells. The results show that three types of SCFAs (acetate, propionate, and butyrate) reduced the production of proinflammatory factors, including TNF-α, IL-1β, IL-6, and NO, and inhibited the vitality of iNOS. Meanwhile, SCFAs enhanced the production of antiinflammatory cytokine IL-10 in lower concentrations (1-1,200 μmol/L). Like NaB, NaAC inhibited LPS-induced NF-κB activation. These results may hold promise on the role that SCFAs have on the prevention and treatment of various inflammatory conditions.
Sigma-Aldrich
Sodium acetate, anhydrous, Molecular Biology, ≥99%
Sigma-Aldrich
Sodium acetate, meets USP testing specifications, anhydrous
Sigma-Aldrich
Sodium acetate, powder, BioReagent, suitable for electrophoresis, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium acetate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium acetate, 99.995% trace metals basis
Sigma-Aldrich
Sodium acetate solution, BioUltra, Molecular Biology, ~3 M in H2O
Sigma-Aldrich
Sodium acetate, anhydrous, BioUltra, suitable for luminescence, Molecular Biology, ≥99.0% (NT)
Sigma-Aldrich
Sodium acetate, puriss. p.a., ACS reagent, reag. Ph. Eur., anhydrous
Sigma-Aldrich
Sodium acetate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium acetate, anhydrous, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium acetate trihydrate, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Sodium acetate trihydrate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium acetate trihydrate, meets USP testing specifications
Sigma-Aldrich
Sodium acetate trihydrate, ACS reagent, ≥99%
Sigma-Aldrich
Sodium acetate trihydrate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium acetate trihydrate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E262, 99.0-101.0% (calc. to the dried substance), ≤0.00002% Al