Merck
  • What happens if people start drinking mouthwash as surrogate alcohol? A quantitative risk assessment.
What happens if people start drinking mouthwash as surrogate alcohol? A quantitative risk assessment.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2012-10-09)
Dirk W Lachenmeier, Yulia B Monakhova, Mariya Markova, Thomas Kuballa, Jürgen Rehm
ABSTRACT
Mouthwash ingestion has been observed in settings of restricted availability to alcoholic beverages such as in hospitals, prisons or military establishments. The literature offers limited evidence that ingredients of mouthwash may have health effects above the effects of ethanol. This study provides a quantitative risk assessment based on analysis of 30 mouthwash samples. All investigated brands contained alcohol, most of them menthol (93%), eucalyptol (90%), benzoic acid (87%), methyl salicylate (67%), and thymol (30%). For low risk drinking scenarios with average levels, only ethanol will exceed acceptable daily intakes (ADI). In worst case scenarios for alcohol dependent consumers ingesting 100g ethanol per day in the form of mouthwash, methyl salicylate will also exceeded the ADI by a factor of 17. The margin of exposure (MOE) for methyl salicylate, benzoates and thymol was below 100 for average scenarios, and below 10 for worst case scenarios, but ethanol is still the most toxic ingredient with MOE below 1. The occasional or even chronic ingestion of mouthwash may not cause health effects except the effects of ethanol alone. Only in extreme exposures, ingredients such as thymol or methyl salicylate could exacerbate the effects of ethanol, especially by contributing to metabolic acidosis.
Product Description
Supelco
Eucalyptol, analytical standard
Sigma-Aldrich
DL-Menthol, ≥95%, FCC, FG
Sigma-Aldrich
(±)-Menthol, racemic, ≥98.0% (GC)
Sigma-Aldrich
Methyl salicylate, ReagentPlus®, ≥99% (GC)
Supelco
Methyl salicylate, analytical standard
Sigma-Aldrich
Methyl salicylate, tested according to Ph. Eur.
Sigma-Aldrich
Methyl salicylate, ≥98%, FCC, FG
Sigma-Aldrich
Eucalyptol, tested according to Ph. Eur.
Sigma-Aldrich
Eucalyptol, natural, ≥99%, FCC, FG
Sigma-Aldrich
Eucalyptol, 99%
Cineole, European Pharmacopoeia (EP) Reference Standard
Supelco
Methyl salicylate, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
1,8-Cineole, primary reference standard
Sigma-Aldrich
Menthol, 99%