Merck
  • Peripheral ostectomy with the use of Carnoy's solution as a rational surgical approach to odontogenic keratocyst: a case report with a 5-year follow-up.
Peripheral ostectomy with the use of Carnoy's solution as a rational surgical approach to odontogenic keratocyst: a case report with a 5-year follow-up.
Vojnosanitetski pregled (2013-02-22)
Stevo Matijević, Zoran Damjanović, Zoran Lazić, Milka Gardasević, Dobrila Radenović-Djurić
ABSTRACT
Odontogenic keratocyst (OKC) is a rare developmental, epithelial and benign cyst of the jaws of odontogenic origin with high recurrence rates. The third molar region, especially the angle of the mandible and the ascending ramus are involved far more frequently than the maxilla. The choice of treatment approach was based on the size of the cyst, recurrence status, and radiographic evidence of cortical perforation. Different surgical treatment options like marsupialization, decompression, enucleation, enucleation with Carnoy's solution, peripheral ostectomy with or without Carnoy's solution, and jaw resection have been discussed in the literature with variable rates of recurrence. We presented a 52-year-old male with orthokeratinized odontogenic keratocyst. Elliptical unilocular radiolucency located in the third molar region and the ascending ramus of the mandible, 40 x 25 mm in diameter with radiographic evidence of cortical perforation at the anterior ramus border of the mandible 20 mm in diameter, was registrated on orthopantomographic radiography. Surgical treatment included enucleation of the cyst and peripheral ostectomy with the use of Carnoy's solution and excision of the overlying attached mucosa. Postoperatively, no paresthesia in the inervation area of the inferior alveolaris nerve was registrated. Recurrences were not registrated within 5 years post-intervention. Treatment of odontogenic keratocyst with enucleation and peripheral ostectomy with the use of Carnoy's solution and excision of the overlying attached mucosa had a very low rate of recurrence. Radical and more aggressive surgical treatments as jaw resection should be reserved for multiple recurrent cysts and when OKC is associated with nevoid basal cell carcinoma syndrome (NBCCS). Following the treatment protocol in the management of OKC and systematic and long-term postsurgical follow-up are considered key elements for successful results.
Sigma-Aldrich
Chloroform, biotech. grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Acetic acid, suitable for luminescence, BioUltra, ≥99.5% (GC)
Supelco
Acetic acid, analytical standard
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, contains amylenes as stabilizer, ACS reagent, ≥99.8%
Sigma-Aldrich
Chloroform, contains ethanol as stabilizer, meets analytical specification of BP, 99-99.4% (GC)
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, ACS spectrophotometric grade, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, contains 100-200 ppm amylenes as stabilizer, ≥99.5%
Sigma-Aldrich
Chloroform, suitable for HPLC, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Supelco
Chloroform, analytical standard
Sigma-Aldrich
Chloroform, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains amylenes as stabilizer
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Chloroform, ReagentPlus®, ≥99.8%, contains 0.5-1.0% ethanol as stabilizer
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Millipore
Bifido Selective Supplement B, suitable for microbiology