Merck
  • Etoposide in combination with low-dose CAG (cytarabine, aclarubicin, G-CSF) for the treatment of relapsed or refractory acute myeloid leukemia: a multicenter, randomized control trial in southwest China.
Etoposide in combination with low-dose CAG (cytarabine, aclarubicin, G-CSF) for the treatment of relapsed or refractory acute myeloid leukemia: a multicenter, randomized control trial in southwest China.
Leukemia research (2013-03-30)
Xi Zhang, Yunlong Li, Yanqi Zhang, Xinghua Chen, Cheng Zhang, Li Gao, Peiyan Kong, Yao Liu, Qin Wen, Yunjing Zeng, Qingyu Wang, Yi Su, Chunsen Wang, Sanbin Wang, Zhong Yuan, Lei Gao
ABSTRACT
In a well-controlled multi-center randomized trial in southwestern China, 228 patients with refractory or relapsed AML were received a low-dose CAG regimen either with etoposide (E-CAG) or without etoposide (CAG). The complete remission (CR) rate, overall survival (OS) and toxicity were evaluated. Patients with E-CAG had a higher CR rate (71.1% vs. CAG 50.9%, P=0.0002). The tolerability appeared to be equivalent. Patients with CR who underwent allogenic hematopoietic stem cell transplantation (allo-HSCT) had a higher five-year OS over those without allo-HSCT (73.8% vs. 10.8%, P=0.000). The E-CAG regimen is expected to become a bridge between relapsed or refractory AML and allo-HSCT.
Sigma-Aldrich
Etoposide, synthetic, 95.0-105.0%, powder
Sigma-Aldrich
Cytosine β-D-arabinofuranoside hydrochloride, crystalline
Sigma-Aldrich
Cytosine β-D-arabinofuranoside, crystalline, ≥90% (HPLC)