Merck
  • A theoretical study of the thermodynamics of microbial growth using Saccharomyces cerevisiae and a different free energy equation.
A theoretical study of the thermodynamics of microbial growth using Saccharomyces cerevisiae and a different free energy equation.
The Quarterly review of biology (2013-08-06)
Edwin H Battley
ABSTRACT
Microbial growth is a biological process that has been previously treated as a chemical reaction operating in accord with the Gibbs free energy equation, Delta G = Delta H-T Delta S. The heat of yeast growth was the first to be measured, in 1856, by direct calorimetry of a large wine vat. Until then there was a tendency for biologists to continue with the old notion that the energy change accompanying the growth of microorganisms was reflected in the amount of heat that was produced during this process. The application of chemical thermodynamics to systems involving microbial growth did not occur until much later. The full application of the Gibbs equation to microbial growth did not take place until the experimental measurement of yeast cell entropy was made in 1997 Further investigations then showed that the quantity of thermal energy for solid substances represented by TS was twice that of the quantity of thermal energy represented by Qab that is experimentally necessary to raise T of a substance from 0/K to T/K. Since there can only be one value for this, the use of the equation Delta X = Delta H-Delta Qab was investigated with respect to microbial growth, and is described in this review.
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Reagent Alcohol, anhydrous, ≤0.003% water
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetic acid, suitable for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-(+)-Glucose, tested according to Ph. Eur.
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC), BioXtra
Supelco
Ethanol-500, 500 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-20 (10 ampules/kit), 20 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-400 (10 ampules/kit), 400 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol Calibration Kit, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-25, 25 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
D-(+)-Glucose, analytical standard
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Supelco
Acetic acid, analytical standard
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Acetic acid solution, suitable for HPLC
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Millipore
Bifido Selective Supplement B, suitable for microbiology
Sigma-Aldrich
L-(−)-Glucose, ≥99%
Sigma-Aldrich
D-(+)-Glucose solution, 100 g/L in H2O, sterile-filtered, BioXtra, suitable for cell culture
Supelco
D-(+)-Glucose solution, 1 mg/mL in 0.1% benzoic acid, standard for enzymatic assay kits GAGO20, GAHK20, STA20, analytical standard