Merck
  • Lauroyl/palmitoyl glycol chitosan gels enhance skin delivery of magnesium ascorbyl phosphate.
Lauroyl/palmitoyl glycol chitosan gels enhance skin delivery of magnesium ascorbyl phosphate.
Journal of cosmetic science (2013-08-13)
Po-Chun Wang, Yan-Ling Huang, Sheng-Shu Hou, Chen-Hsi Chou, Jui-Chen Tsai
ABSTRACT
Palmitoyl glycol chitosan (GCP) hydrogel has been reported as erodible controlled-release systems for the delivery of both hydrophilic and hydrophobic molecules. In this study we prepared lauroyl/palmitoyl glycol chitosan (GCL/GCP) in gel form and evaluated their application for skin delivery of the hydrophilic compound, magnesium ascorbyl phosphate (MAP), which is widely used in cosmetic formulations. Release of MAP from the polymer gels was significantly decreased with increasing concentration of GCL/GCP in the formulations in comparison with glycol chitosan (GC). In both aqueous and 10% ethanol vehicles, MAP flux was increased 1.58- to 3.96-fold of 1% GC from 1% GCL/GCP. Increase in MAP flux was correlated to the increase in GCL/GCP concentration prepared in 10% ethanol vehicle. GCL/GCP, in either water or 10% ethanol vehicles, increased the skin penetration and skin deposition of MAP in comparison with GC, hydroxypropylmethylcellulose, and carbopol, while sustaining its release from the polymer gels. Both the enhancement in skin penetration/deposition and sustained release of MAP were depended on polymer concentration. Also, with increase in polymer concentration, epidermal to dermal drug deposition ratio tended to increase, which will be beneficial to its activity in the epidermis, such as inhibition of tyrosinase and protection from UV damage. These data suggested both GCL and GCP can be applied as delivery vehicles to improve percutaneous absorption of MAP.
Sigma-Aldrich
Chitosan, high molecular weight
Supelco
Sodium ascorbate, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Ascorbic acid, analytical standard
Sigma-Aldrich
(+)-Sodium L-ascorbate, BioXtra, ≥99.0% (NT)
Sigma-Aldrich
(+)-Sodium L-ascorbate, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
L-Ascorbic acid, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
L-Ascorbic acid, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
L-Ascorbic acid, ACS reagent, ≥99%
Sigma-Aldrich
L-Ascorbic acid, reagent grade, crystalline
Sigma-Aldrich
L-Ascorbic acid, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
L-Ascorbic acid, reagent grade
Sigma-Aldrich
L-Ascorbic acid, 99%
Sigma-Aldrich
L-Ascorbic acid, puriss. p.a., ≥99.0% (RT)
Sigma-Aldrich
L-Ascorbic acid, BioUltra, ≥99.5% (RT)
Sigma-Aldrich
L-Ascorbic acid, meets USP testing specifications
Supelco
L-Ascorbic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Ascorbic acid, tested according to Ph. Eur.
Sigma-Aldrich
L-Ascorbic acid, FCC, FG
Sigma-Aldrich
Chitosan, from shrimp shells, practical grade
Sigma-Aldrich
Chitosan, low molecular weight
Sigma-Aldrich
Chitosan, from shrimp shells, ≥75% (deacetylated)
Sigma-Aldrich
Chitosan, medium molecular weight
Sigma-Aldrich
(+)-Sodium L-ascorbate, crystalline, ≥98%
Supelco
Ascorbic Acid, Pharmaceutical Secondary Standard; Certified Reference Material
Ascorbic acid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Chitosan from shrimp shells, low-viscous