Merck
  • The pan-ErbB receptor tyrosine kinase inhibitor canertinib promotes apoptosis of malignant melanoma in vitro and displays anti-tumor activity in vivo.
The pan-ErbB receptor tyrosine kinase inhibitor canertinib promotes apoptosis of malignant melanoma in vitro and displays anti-tumor activity in vivo.
Biochemical and biophysical research communications (2011-10-11)
Emelie A Djerf Severinsson, Cecilia Trinks, Henrik Gréen, Avni Abdiu, Anna-Lotta Hallbeck, Olle Stål, Thomas M Walz
ABSTRACT
The ErbB receptor family has been suggested to constitute a therapeutic target for tumor-specific treatment of malignant melanoma. Here we investigate the effect of the pan-ErbB tyrosine kinase inhibitor canertinib on cell growth and survival in human melanoma cells in vitro and in vivo. Canertinib significantly inhibited growth of cultured melanoma cells, RaH3 and RaH5, in a dose-dependent manner as determined by cell counting. Half-maximum growth inhibitory dose (IC(50)) was approximately 0.8 μM and by 5 μM both cell lines were completely growth-arrested within 72 h of treatment. Incubation of exponentially growing RaH3 and RaH5 with 1 μM canertinib accumulated the cells in the G(1)-phase of the cell cycle within 24h of treatment without induction of apoptosis as determined by flow cytometry. Immunoblot analysis showed that 1 μM canertinib inhibited ErbB1-3 receptor phosphorylation with a concomitant decrease of Akt-, Erk1/2- and Stat3 activity in both cell lines. In contrast to the cytostatic effect observed at doses ≤ 5μM canertinib, higher concentrations induced apoptosis as demonstrated by the Annexin V method and Western blot analysis of PARP cleavage. Furthermore, canertinib significantly inhibited growth of RaH3 and RaH5 melanoma xenografts in nude mice. Pharmacological targeting of the ErbB receptors may prove successful in the treatment of patients with metastatic melanoma.
Sigma-Aldrich
CI-1033, ≥98% (HPLC)
Pricing and availability is not currently available.