Merck
Selfish DNA: a pharmaceutical perspective.
Die Pharmazie (2013-08-09)
T Winckler
ABSTRACT
Almost 25 years ago, Theo Dingermann published the discovery of a new mobile genetic element in the unicellular microbe Dictyostelium discoideum in the journal Science. An interesting property of this new molecular parasite, the Dictyostelium Repetitive Element (DRE), was that all integrations were found approximately 50 base pairs (bp) upstream of transfer RNA (tRNA) genes in the D. discoideum genome, thus implying an active targeting mechanism to avoid the disruption of host cell genes by the retrotransposition process. Since then, the facultative multicellular "social amoeba" D. discoideum has become a popular model for analyzing complex cellular functions such as cell movement, chemotaxis, phagocytosis, and cell differentiation, important areas of biomedical research that are often hard to investigate in cells from "higher organisms" including humans. Therefore, progress in the development of methods to study Dictyostelium biology has also provoked research on transposable elements in this organism. Early work on the DRE element suggested that studying its molecular mechanism of site-specific integration might promote human gene therapy technology through the design of integrating gene transfer vectors with low intrinsic genotoxic potential. In this review article, I will briefly review the original research performed on the DRE transposable element in the Dingermann lab and report on how the emergence of genomics technologies and the development of tools to analyze de novo retrotransposition events in D. discoideum cells will expand our knowledge of DRE biology in the future.
Sigma-Aldrich
Ribonucleic acid, transfer from Escherichia coli, Type XX, Strain W, lyophilized powder
Sigma-Aldrich
Ribonucleic acid, transfer from bovine liver, Type XI, lyophilized powder
Sigma-Aldrich
Ribonucleic acid, transfer from baker′s yeast (S. cerevisiae), buffered aqueous solution
Sigma-Aldrich
Ribonucleic acid, transfer from baker′s yeast (S. cerevisiae), Type X-SA, lyophilized powder
Sigma-Aldrich
Ribonucleic acid, transfer from baker′s yeast (S. cerevisiae), buffered aqueous solution
Sigma-Aldrich
Ribonucleic acid, transfer from wheat germ, Type V, 15-19 units/mg solid