Merck
Diagnostic use of cerebral and extracerebral oxysterols.
Clinical chemistry and laboratory medicine (2004-04-06)
Valerio Leoni, Thomas Masterman, Fariba S Mousavi, Bengt Wretlind, Lars-Olof Wahlund, Ulf Diczfalusy, Jan Hillert, Ingemar Björkhem
ABSTRACT
24S-Hydroxycholesterol (24OHC) and 27-hydroxycholesterol (27OHC) are two structurally similar oxysterols of different origins--the former almost exclusively formed in the brain and the latter formed to a lesser extent in the brain than in most other organs. HYPOTHESIS TO BE TESTED: Neuronal damage and/or demyelination causes increased flux of 24OHC from the brain into the cerebrospinal fluid (CSF), whereas a defect blood-brain barrier causes increased flux of 27OHC from the circulation into the CSF. Isotope dilution-mass spectrometry was used to assay the two oxysterols in CSF and plasma from more than 250 patients with different neurological and geriatric diseases. The CSF-levels of the two oxysterols were much more affected by the different diseases than the plasma levels. Patients with active demyelinating diseases had increased levels of 24OHC in CSF with a relatively high 24OHC/27OHC ratio. Patients with meningitis in general had high levels of both steroids with a low 24OHC/27OHC ratio. Patients with Alzheimer's disease had slightly increased levels of 24OHC in CSF with less increase in 27OHC. Patients with multiple sclerosis had a tendency to have higher levels of 24OHC during active periods with a high 24OHC/ 27OHC ratio. Measurements of the two oxysterols in CSF and plasma may add significantly to existing biochemical methods for evaluation of neurological diseases.
Sigma-Aldrich
Nickel, powder, <1 μm, 99.8% trace metals basis
Supelco
L-Cysteine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Glutamic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Glutamine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Asparagine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Isoleucine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Millipore
Bifido Selective Supplement B, suitable for microbiology
Sigma-Aldrich
L-Serine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Bismuth, granular, ≥99.99% trace metals basis
Sigma-Aldrich
L-Isoleucine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Tungsten, foil, thickness 0.127 mm, ≥99.9% trace metals basis
Sigma-Aldrich
L-Glutamic acid, FCC
Sigma-Aldrich
Calcium, dendritic pieces, purified by distillation, 99.99% trace metals basis
Sigma-Aldrich
Pyruvic acid, ≥97%, FG
Sigma-Aldrich
Tin, foil, thickness 0.5 mm, 99.998% trace metals basis
Sigma-Aldrich
Copper, powder, 99.99% trace metals basis
Sigma-Aldrich
Molybdenum, wire, diam. 1.0 mm, 99.95% trace metals basis
Sigma-Aldrich
Calcium, granular, 99%
Sigma-Aldrich
L-Cysteine, 97%
Sigma-Aldrich
Titanium, foil, thickness 0.5 mm, 99.99% trace metals basis
Sigma-Aldrich
Succinic acid, ACS reagent, ≥99.0%
Sigma-Aldrich
Tungsten, wire, diam. 0.5 mm, ≥99.9% trace metals basis
Sigma-Aldrich
L-Cysteine, ≥97%, FG
Sigma-Aldrich
Tin, powder, <45 μm particle size, 99.8% trace metals basis
Sigma-Aldrich
Calcium, turnings, 99% trace metals basis
Sigma-Aldrich
Copper, powder, <425 μm, 99.5% trace metals basis
Sigma-Aldrich
Titanium, powder, <45 μm avg. part. size, 99.98% trace metals basis
Sigma-Aldrich
Copper, powder (dendritic), <45 μm, 99.7% trace metals basis
Sigma-Aldrich
Copper, foil, thickness 0.25 mm, 99.98% trace metals basis
Sigma-Aldrich
Methylmalonic acid, 99%