Merck
  • Prediction of skeletal muscle and fat mass in patients with advanced cancer using a metabolomic approach.
Prediction of skeletal muscle and fat mass in patients with advanced cancer using a metabolomic approach.
The Journal of nutrition (2011-12-14)
Cynthia Stretch, Thomas Eastman, Rupasri Mandal, Roman Eisner, David S Wishart, Marina Mourtzakis, Carla M M Prado, Sambasivarao Damaraju, Ronald O Ball, Russell Greiner, Vickie E Baracos
ABSTRACT
Urine and plasma metabolites originate from endogenous metabolic pathways in different organs and exogenous sources (diet). Urine and plasma were obtained from advanced cancer patients and investigated to determine if variations in lean and fat mass, dietary intake, and energy metabolism relate to variation in metabolite profiles. Patients (n = 55) recorded their diets for 3 d and after an overnight fast they were evaluated by DXA and indirect calorimetry. Metabolites were measured by NMR and direct injection MS. Three algorithms were used [partial least squares discriminant-analysis, support vector machines (SVM), and least absolute shrinkage and selection operator] to relate patients' plasma/urine metabolic profile with their dietary/physiological assessments. Leave-one-out cross-validation and permutation testing were conducted to determine statistical validity. None of the algorithms, using 63 urine metabolites, could learn to predict variations in individual's resting energy expenditure, respiratory quotient, or their intake of total energy, fat, sugar, or carbohydrate. Urine metabolites predicted appendicular lean tissue (skeletal muscle) with excellent cross-validation accuracy (98% using SVM). Total lean tissue correlated highly with appendicular muscle (Pearson r = 0.98; P < 0.0001) and gave similar cross-validation accuracies. Fat mass was effectively predicted using the 63 urine metabolites or the 143 plasma metabolites, exclusively. In conclusion, in this population, lean and fat mass variation could be effectively predicted using urinary metabolites, suggesting a potential role for metabolomics in body composition research. Furthermore, variation in lean and fat mass potentially confounds metabolomic studies attempting to characterize diet or disease conditions. Future studies should account or correct for such variation.
Product Description
Supelco
Glycine, analytical standard, for nitrogen determination according to Kjeldahl method
Supelco
L-Valine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Glycine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
D-Mannitol, ≥99.9999% (metals basis), for boron determination
Supelco
Glycine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Ethanol-25, 25 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Mannitol, European Pharmacopoeia (EP) Reference Standard
Supelco
Ethanol-400 (10 ampules/kit), 400 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-500, 500 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Ethanol-20 (10 ampules/kit), 20 mg/dL in H2O, ampule of 10 × 1.2 mL, certified reference material, Cerilliant®
Supelco
Mannitol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Betaine solution, 5 M, PCR Reagent
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
L-Valine, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Valine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
D-Mannitol, meets EP, FCC, USP testing specifications
Sigma-Aldrich
D-Mannitol, ≥98% (GC)
Sigma-Aldrich
D-Mannitol, BioXtra, ≥98% (HPLC)
Sigma-Aldrich
cis-Aconitic acid, ≥98%
Sigma-Aldrich
D-Mannitol, ACS reagent
Sigma-Aldrich
D-Mannitol, ≥98% (GC), suitable for plant cell culture
SAFC
L-Valine
SAFC
Glycine
Sigma-Aldrich
D-Mannitol, tested according to Ph. Eur.
Sigma-Aldrich
Acetic acid, suitable for luminescence, BioUltra, ≥99.5% (GC)
Supelco
Acetic acid, analytical standard
Sigma-Aldrich
Glycine, BioUltra, Molecular Biology, ≥99.0% (NT)
Sigma-Aldrich
L-Valine, BioUltra, ≥99.5% (NT)