Merck
  • Occurrence and distribution of pharmaceutically active and endocrine disrupting compounds in Singapore's marine environment: influence of hydrodynamics and physical-chemical properties.
Occurrence and distribution of pharmaceutically active and endocrine disrupting compounds in Singapore's marine environment: influence of hydrodynamics and physical-chemical properties.
Environmental pollution (Barking, Essex : 1987) (2013-07-31)
Stéphane Bayen, Hui Zhang, Malan Manish Desai, Seng Keat Ooi, Barry C Kelly
ABSTRACT
The fate and exposure risks of pharmaceutically active compounds (PhACs) and endocrine disrupting chemicals (EDCs) in marine environments are not well-understood. In this study we developed a multi-residue analytical method for quantifying concentrations of forty target compounds in seawater from Singapore. Analyses of samples (n = 24) from eight sites showed the occurrence of several compounds, including gemfibrozil (<0.09-19.8 ng/L), triclosan (<0.55-10.5 ng/L), carbamazepine (<0.28-10.9 ng/L) and ibuprofen (<2.2-9.1 ng/L). A 3D hydrodynamic model for Singapore was used to predict residence time (tR). Principal Components Analysis revealed a strong relationship between tR and contaminant concentrations. While source emissions are undoubtedly important, proximate distance to a wastewater treatment plant had little influence on concentrations. The site with the greatest tR, which exhibited the highest concentrations, is adjacent to Singapore's largest protected wetland reserve. The results highlight an important linkage between hydrodynamic behavior and contaminant exposure risks in complex coastal marine ecosystems.
Product Description
Ibuprofen, European Pharmacopoeia (EP) Reference Standard
Supelco
Carbamazepine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Ibuprofen, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Carbamazepine, powder
Sigma-Aldrich
Carbamazepine, meets USP testing specifications
Supelco
Ibuprofen
Sigma-Aldrich
Ibuprofen, meets USP testing specifications
Sigma-Aldrich
Ibuprofen, ≥98% (GC)
Supelco
Ibuprofen sodium salt, ≥98% (GC)
Ibuprofen for peak identification, European Pharmacopoeia (EP) Reference Standard
Gemfibrozil for system suitability, European Pharmacopoeia (EP) Reference Standard
Supelco
Ibuprofen solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Gemfibrozil, Pharmaceutical Secondary Standard; Certified Reference Material
Gemfibrozil, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
(S)-(+)-Ibuprofen, ReagentPlus®, 99%
Sigma-Aldrich
Triclosan, 97.0-103.0% (active substance, GC)
Supelco
Carbamazepine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Gemfibrozil