Merck
  • Monanchomycalin C, a new pentacyclic guanidine alkaloid from the far-eastern marine sponge Monanchora pulchra.
Monanchomycalin C, a new pentacyclic guanidine alkaloid from the far-eastern marine sponge Monanchora pulchra.
Natural product communications (2013-12-21)
Ksenya M Tabakmakher, Vladimir A Denisenko, Alla G Guzii, Pavel S Dmitrenok, Sergey A Dyshlovoy, Hyi-Seung Lee, Tatyana N Makarieva
ABSTRACT
A new pentacyclic guanidine alkaloid, monanchomycalin C (1), along with the earlier known ptilomycalin A (2), were isolated from the Far-Eastern marine sponge Monanchora pulchra. The structure of 1 was elucidated using 1D and 2D NMR spectroscopic and ma ss spectrometric da ta. Compounds 1 and 2 exhibited cytotoxic activities against human breast cancer MDA-MB-231 cells with IC50 values of 8.2 microM and 4.3 pM, respectively.
Sigma-Aldrich
Guanidine hydrochloride, organic base and chaeotropic agent, ≥99% (titration)
Sigma-Aldrich
Guanidine hydrochloride, Molecular Biology, ≥99%
Sigma-Aldrich
Guanidine hydrochloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Guanidine hydrochloride, ≥98%
Sigma-Aldrich
Guanidine hydrochloride, ≥99.0% (AT)
Sigma-Aldrich
Guanidine sulfate salt, 99%
Sigma-Aldrich
Guanidine hydrochloride solution, Colorless liquid, 7.8 - 8.3 M, pH- 4.5 - 5.5
Sigma-Aldrich
Guanidine hydrochloride solution, BioUltra, ~8 M in H2O