Merck
  • Pharmacology and pathophysiology of mutated KCNJ5 found in adrenal aldosterone-producing adenomas.
Pharmacology and pathophysiology of mutated KCNJ5 found in adrenal aldosterone-producing adenomas.
Endocrinology (2014-02-11)
P Tauber, D Penton, J Stindl, E Humberg, I Tegtmeier, C Sterner, F Beuschlein, M Reincke, J Barhanin, S Bandulik, R Warth
ABSTRACT
Somatic mutations of the potassium channel KCNJ5 are found in 40% of aldosterone producing adenomas (APAs). APA-related mutations of KCNJ5 lead to a pathological Na(+) permeability and a rise in cytosolic Ca(2+), the latter presumably by depolarizing the membrane and activating voltage-gated Ca(2+) channels. The aim of this study was to further investigate the effects of mutated KCNJ5 channels on intracellular Na(+) and Ca(2+) homeostasis in human adrenocortical NCI-H295R cells. Expression of mutant KCNJ5 led to a 2-fold increase in intracellular Na(+) and, in parallel, to a substantial rise in intracellular Ca(2+). The increase in Ca(2+) appeared to be caused by activation of voltage-gated Ca(2+) channels and by an impairment of Ca(2+) extrusion by Na(+)/Ca(2+) exchangers. The mutated KCNJ5 exhibited a pharmacological profile that differed from the one of wild-type channels. Mutated KCNJ5 was less Ba(2+) and tertiapin-Q sensitive but was inhibited by blockers of Na(+) and Ca(2+)-transporting proteins, such as verapamil and amiloride. The clinical use of these drugs might influence aldosterone levels in APA patients with KCNJ5 mutations. This might implicate diagnostic testing of APAs and could offer new therapeutic strategies.
Product Description
Supelco
Lidocaine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
(±)-Verapamil hydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Amiloride hydrochloride hydrate, ≥98% (HPLC), powder
Sigma-Aldrich
(±)-Verapamil hydrochloride, ≥99% (titration), powder
Sigma-Aldrich
Lidocaine, powder
Sigma-Aldrich
Lidocaine, analytical standard
Supelco
Flufenamic acid, analytical standard, for drug analysis
Sigma-Aldrich
Nifedipine, ≥98% (HPLC), powder
Nifedipine, European Pharmacopoeia (EP) Reference Standard
Supelco
Verapamil hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
Lidocaine, European Pharmacopoeia (EP) Reference Standard
USP
Nifedipine, United States Pharmacopeia (USP) Reference Standard
USP
Lidocaine, United States Pharmacopeia (USP) Reference Standard
Supelco
Nifedipine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Barium, beads, 0.5-2.0 mm particle size, 99% trace metals basis
Supelco
Methanesulfonic acid, suitable for HPLC, LiChropur, ≥99.5% (T)
Verapamil hydrochloride, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium, 99.95% trace metals basis, ingot
Sigma-Aldrich
Sodium, ACS reagent, dry
Sigma-Aldrich
Sodium, cubes, contains mineral oil, 99.9% trace metals basis
Sigma-Aldrich
Sodium, 25-35 wt % dispersion in paraffin
Sigma-Aldrich
Barium, dendritic pieces, purified by distillation, 99.99% trace metals basis
Sigma-Aldrich
Methanesulfonic acid solution, 70 wt. % in H2O
Sigma-Aldrich
Barium, pieces, 1 cm, 99% trace metals basis
Sigma-Aldrich
Methanesulfonic acid solution, 4 M (with 0.2% (w/v) tryptamine)
Sigma-Aldrich
5-(N-Ethyl-N-isopropyl)amiloride
Sigma-Aldrich
Barium, rod, diam. ~2 cm, ≥99% trace metals basis
Sigma-Aldrich
Sodium, in kerosene, pieces (large), ≥99.8% (sodium basis)
Sigma-Aldrich
Methanesulfonic acid, ≥99.0%
USP
Verapamil hydrochloride, United States Pharmacopeia (USP) Reference Standard